Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15111553-15111824 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYDE1
Entrez Gene ID
85360
Gene Name
synapse defective Rho GTPase homolog 1
Gene Aliases
7h3, SYD1
UniGene
Hs.528701
Gene Chromosome Location
Chr.19: 15107364-15114988 on Build GRCh38
Transcripts
NM_001300910.1 NM_033025.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5772951 c.1425C>T p.L475L Chr.19: 15111639-15111639 on Build GRCh38
6277596 c.1432T>A p.Y478N Chr.19: 15111646-15111646 on Build GRCh38
4885129 c.1446G>T p.L482F Chr.19: 15111660-15111660 on Build GRCh38
1680745 c.1438C>T p.R480* Chr.19: 15111652-15111652 on Build GRCh38
5388855 c.1526C>T p.P509L Chr.19: 15111740-15111740 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374991037 Chr.19: 15111700-15111700 on Build GRCh38
rs141830780 Chr.19: 15111716-15111716 on Build GRCh38
rs772266058 Chr.19: 15111750-15111750 on Build GRCh38
rs33980637 Chr.19: 15111681-15111681 on Build GRCh38
rs1048270287 Chr.19: 15111737-15111737 on Build GRCh38
rs761003994 Chr.19: 15111627-15111627 on Build GRCh38
rs773462650 Chr.19: 15111618-15111618 on Build GRCh38
rs184293589 Chr.19: 15111758-15111758 on Build GRCh38
rs745525231 Chr.19: 15111715-15111715 on Build GRCh38
rs748288079 Chr.19: 15111740-15111740 on Build GRCh38
rs181180550 Chr.19: 15111611-15111611 on Build GRCh38
rs755423646 Chr.19: 15111622-15111622 on Build GRCh38
rs1023024892 Chr.19: 15111725-15111728 on Build GRCh38
rs771003925 Chr.19: 15111763-15111763 on Build GRCh38
rs779170644 Chr.19: 15111667-15111667 on Build GRCh38
rs2040858 Chr.19: 15111617-15111617 on Build GRCh38
rs760957685 Chr.19: 15111760-15111760 on Build GRCh38
rs766761902 Chr.19: 15111645-15111645 on Build GRCh38
rs770943850 Chr.19: 15111736-15111736 on Build GRCh38
rs979243434 Chr.19: 15111575-15111575 on Build GRCh38
rs776881144 Chr.19: 15111745-15111747 on Build GRCh38
rs756735660 Chr.19: 15111693-15111693 on Build GRCh38
rs558734014 Chr.19: 15111734-15111734 on Build GRCh38
rs748984741 Chr.19: 15111718-15111718 on Build GRCh38
rs774399408 Chr.19: 15111738-15111738 on Build GRCh38
rs113617557 Chr.19: 15111630-15111630 on Build GRCh38
rs943943445 Chr.19: 15111705-15111705 on Build GRCh38
rs751103685 Chr.19: 15111687-15111687 on Build GRCh38
rs576323020 Chr.19: 15111658-15111658 on Build GRCh38
rs761942935 Chr.19: 15111600-15111600 on Build GRCh38
rs755526774 Chr.19: 15111668-15111668 on Build GRCh38

View Full Product Details