Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 154543884-154544127 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TDRD10
Entrez Gene ID
126668
Gene Name
tudor domain containing 10
Gene Aliases
-
UniGene
Hs.387671
Gene Chromosome Location
Chr.1: 154502219-154548147 on Build GRCh38
Transcripts
NM_001098475.1 NM_182499.3 XM_006711156.2 XM_011509152.1 XM_011509153.1 XM_011509154.1 XM_011509155.1 XM_011509156.1 XM_011509157.1 XM_011509158.2 XM_011509159.2 XM_011509160.2 XM_011509161.1 XM_017000255.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4997885 c.643G>A p.V215I Chr.1: 154544102-154544102 on Build GRCh38
6343181 c.519G>T p.V173V Chr.1: 154543978-154543978 on Build GRCh38
1334446 c.573C>T p.S191S Chr.1: 154544032-154544032 on Build GRCh38
3474700 c.621C>T p.T207T Chr.1: 154544080-154544080 on Build GRCh38
3802030 c.649G>A p.E217K Chr.1: 154544108-154544108 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201320017 Chr.1: 154544015-154544015 on Build GRCh38
rs143192137 Chr.1: 154544033-154544033 on Build GRCh38
rs147377261 Chr.1: 154543981-154543981 on Build GRCh38
rs757765828 Chr.1: 154544074-154544074 on Build GRCh38
rs373556830 Chr.1: 154543959-154543959 on Build GRCh38
rs766356300 Chr.1: 154544036-154544036 on Build GRCh38
rs546615227 Chr.1: 154544061-154544061 on Build GRCh38
rs142719065 Chr.1: 154543916-154543916 on Build GRCh38
rs200051309 Chr.1: 154543979-154543979 on Build GRCh38
rs1019770946 Chr.1: 154543976-154543976 on Build GRCh38
rs780852581 Chr.1: 154544025-154544025 on Build GRCh38
rs760663272 Chr.1: 154543945-154543945 on Build GRCh38
rs6678164 Chr.1: 154544095-154544095 on Build GRCh38
rs751346307 Chr.1: 154543953-154543953 on Build GRCh38
rs375653959 Chr.1: 154544040-154544040 on Build GRCh38
rs956944081 Chr.1: 154543915-154543915 on Build GRCh38
rs780704841 Chr.1: 154543958-154543958 on Build GRCh38
rs780360116 Chr.1: 154544083-154544083 on Build GRCh38
rs759988293 Chr.1: 154544000-154544000 on Build GRCh38
rs769568577 Chr.1: 154544046-154544046 on Build GRCh38
rs758963449 Chr.1: 154543910-154543910 on Build GRCh38
rs778294717 Chr.1: 154543967-154543967 on Build GRCh38
rs3811448 Chr.1: 154544102-154544102 on Build GRCh38
rs907124847 Chr.1: 154543902-154543902 on Build GRCh38
rs775640036 Chr.1: 154543941-154543941 on Build GRCh38
rs957045494 Chr.1: 154544042-154544042 on Build GRCh38
rs746297703 Chr.1: 154544080-154544080 on Build GRCh38
rs774655817 Chr.1: 154543917-154543917 on Build GRCh38
rs753287462 Chr.1: 154544069-154544069 on Build GRCh38
rs371972287 Chr.1: 154543969-154543969 on Build GRCh38
rs76289529 Chr.1: 154543928-154543928 on Build GRCh38
rs779384867 Chr.1: 154544077-154544077 on Build GRCh38
rs201077832 Chr.1: 154544023-154544023 on Build GRCh38
rs750360703 Chr.1: 154543949-154543949 on Build GRCh38
rs758375424 Chr.1: 154543950-154543950 on Build GRCh38
rs532940684 Chr.1: 154544062-154544062 on Build GRCh38
rs12750774 Chr.1: 154544001-154544001 on Build GRCh38
rs751439221 Chr.1: 154544037-154544037 on Build GRCh38
rs887176880 Chr.1: 154543956-154543956 on Build GRCh38
rs766303059 Chr.1: 154543951-154543951 on Build GRCh38
rs368379318 Chr.1: 154544068-154544068 on Build GRCh38
rs745982586 Chr.1: 154544047-154544047 on Build GRCh38
rs369017708 Chr.1: 154544047-154544047 on Build GRCh38
rs760855348 Chr.1: 154544020-154544020 on Build GRCh38
rs901377443 Chr.1: 154544034-154544034 on Build GRCh38
rs539002486 Chr.1: 154544082-154544082 on Build GRCh38
rs183584721 Chr.1: 154543929-154543929 on Build GRCh38
rs771284999 Chr.1: 154543972-154543972 on Build GRCh38
rs201832003 Chr.1: 154544032-154544032 on Build GRCh38
rs755559844 Chr.1: 154543966-154543966 on Build GRCh38
rs114098415 Chr.1: 154543946-154543946 on Build GRCh38
rs772400586 Chr.1: 154544014-154544014 on Build GRCh38
rs774532547 Chr.1: 154544030-154544030 on Build GRCh38

View Full Product Details