Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 133401705-133401966 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STKLD1
Entrez Gene ID
169436
Gene Name
serine/threonine kinase like domain containing 1
Gene Aliases
C9orf96, SgK071, Sk521
UniGene
Hs.720195
Gene Chromosome Location
Chr.9: 133376413-133406096 on Build GRCh38
Transcripts
NM_153710.4 NR_103997.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3213437 c.1278delC p.E428fs*13 Chr.9: 133401817-133401817 on Build GRCh38
6617055 c.1199-2A>C p.? Chr.9: 133401736-133401736 on Build GRCh38
422465 c.1228C>T p.P410S Chr.9: 133401767-133401767 on Build GRCh38
5776742 c.1281C>T p.P427P Chr.9: 133401820-133401820 on Build GRCh38
5520337 c.1226C>T p.A409V Chr.9: 133401765-133401765 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782576495 Chr.9: 133401843-133401843 on Build GRCh38
rs782764801 Chr.9: 133401902-133401902 on Build GRCh38
rs781815220 Chr.9: 133401743-133401743 on Build GRCh38
rs781877634 Chr.9: 133401773-133401773 on Build GRCh38
rs978105450 Chr.9: 133401881-133401881 on Build GRCh38
rs782686532 Chr.9: 133401755-133401755 on Build GRCh38
rs782176295 Chr.9: 133401780-133401780 on Build GRCh38
rs782543174 Chr.9: 133401754-133401754 on Build GRCh38
rs150111949 Chr.9: 133401833-133401833 on Build GRCh38
rs781828624 Chr.9: 133401833-133401835 on Build GRCh38
rs781909747 Chr.9: 133401821-133401821 on Build GRCh38
rs782086356 Chr.9: 133401918-133401918 on Build GRCh38
rs782630938 Chr.9: 133401857-133401857 on Build GRCh38
rs138146001 Chr.9: 133401868-133401868 on Build GRCh38
rs782387366 Chr.9: 133401760-133401760 on Build GRCh38
rs939475639 Chr.9: 133401739-133401739 on Build GRCh38
rs782074352 Chr.9: 133401817-133401817 on Build GRCh38
rs782508745 Chr.9: 133401828-133401828 on Build GRCh38
rs150926353 Chr.9: 133401784-133401784 on Build GRCh38
rs28418384 Chr.9: 133401839-133401839 on Build GRCh38
rs782247289 Chr.9: 133401759-133401759 on Build GRCh38
rs782292872 Chr.9: 133401789-133401789 on Build GRCh38
rs909640566 Chr.9: 133401941-133401944 on Build GRCh38
rs968424294 Chr.9: 133401879-133401879 on Build GRCh38
rs782182852 Chr.9: 133401882-133401882 on Build GRCh38
rs200005667 Chr.9: 133401861-133401861 on Build GRCh38
rs140796131 Chr.9: 133401820-133401820 on Build GRCh38
rs782172186 Chr.9: 133401897-133401897 on Build GRCh38
rs782074801 Chr.9: 133401793-133401793 on Build GRCh38
rs150258727 Chr.9: 133401853-133401853 on Build GRCh38
rs782561000 Chr.9: 133401782-133401782 on Build GRCh38
rs1028516530 Chr.9: 133401885-133401885 on Build GRCh38
rs763668159 Chr.9: 133401765-133401765 on Build GRCh38
rs905719073 Chr.9: 133401802-133401802 on Build GRCh38
rs782335607 Chr.9: 133401874-133401874 on Build GRCh38
rs186805420 Chr.9: 133401937-133401937 on Build GRCh38
rs201751407 Chr.9: 133401738-133401738 on Build GRCh38
rs138721622 Chr.9: 133401836-133401836 on Build GRCh38
rs782332667 Chr.9: 133401883-133401883 on Build GRCh38
rs587604470 Chr.9: 133401725-133401725 on Build GRCh38
rs782737798 Chr.9: 133401921-133401921 on Build GRCh38
rs781926399 Chr.9: 133401809-133401809 on Build GRCh38
rs782278720 Chr.9: 133401849-133401849 on Build GRCh38
rs200615951 Chr.9: 133401911-133401911 on Build GRCh38
rs1056627436 Chr.9: 133401757-133401757 on Build GRCh38
rs781993288 Chr.9: 133401790-133401790 on Build GRCh38
rs1056767232 Chr.9: 133401930-133401930 on Build GRCh38
rs782020704 Chr.9: 133401895-133401895 on Build GRCh38
rs781963015 Chr.9: 133401875-133401875 on Build GRCh38
rs781956257 Chr.9: 133401891-133401891 on Build GRCh38

View Full Product Details