Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 44157134-44157363 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF234
Entrez Gene ID
10780
Gene Name
zinc finger protein 234
Gene Aliases
HZF4, ZNF269
UniGene
Hs.235992
Gene Chromosome Location
Chr.19: 44141530-44160309 on Build GRCh38
Transcripts
NM_001144824.1 NM_006630.2 XM_006722974.3 XM_017026149.1 XM_017026150.1 XM_017026151.1 XM_017026152.1 XM_017026153.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4079087 c.1147T>C p.Y383H Chr.19: 44157163-44157163 on Build GRCh38
3535459 c.1171C>A p.Q391K Chr.19: 44157187-44157187 on Build GRCh38
5707069 c.1315C>T p.Q439* Chr.19: 44157331-44157331 on Build GRCh38
5329478 c.1173G>T p.Q391H Chr.19: 44157189-44157189 on Build GRCh38
3535460 c.1264C>T p.H422Y Chr.19: 44157280-44157280 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777846508 Chr.19: 44157200-44157200 on Build GRCh38
rs757293878 Chr.19: 44157229-44157229 on Build GRCh38
rs771820073 Chr.19: 44157276-44157276 on Build GRCh38
rs765783852 Chr.19: 44157271-44157271 on Build GRCh38
rs748108043 Chr.19: 44157248-44157248 on Build GRCh38
rs939587528 Chr.19: 44157313-44157313 on Build GRCh38
rs200729739 Chr.19: 44157302-44157302 on Build GRCh38
rs201556875 Chr.19: 44157274-44157274 on Build GRCh38
rs1045342875 Chr.19: 44157222-44157222 on Build GRCh38
rs756548547 Chr.19: 44157187-44157187 on Build GRCh38
rs759074594 Chr.19: 44157279-44157279 on Build GRCh38
rs768526277 Chr.19: 44157238-44157238 on Build GRCh38
rs756460513 Chr.19: 44157299-44157299 on Build GRCh38
rs749434109 Chr.19: 44157207-44157207 on Build GRCh38
rs767933259 Chr.19: 44157284-44157284 on Build GRCh38
rs757376680 Chr.19: 44157317-44157317 on Build GRCh38
rs746024802 Chr.19: 44157237-44157237 on Build GRCh38
rs779241155 Chr.19: 44157230-44157230 on Build GRCh38
rs769908534 Chr.19: 44157253-44157253 on Build GRCh38
rs753005346 Chr.19: 44157290-44157290 on Build GRCh38
rs778814776 Chr.19: 44157323-44157323 on Build GRCh38
rs773279486 Chr.19: 44157262-44157262 on Build GRCh38
rs974278081 Chr.19: 44157285-44157285 on Build GRCh38
rs754318858 Chr.19: 44157310-44157310 on Build GRCh38
rs376364041 Chr.19: 44157329-44157329 on Build GRCh38
rs753238363 Chr.19: 44157185-44157185 on Build GRCh38
rs929833064 Chr.19: 44157309-44157309 on Build GRCh38
rs372837361 Chr.19: 44157263-44157263 on Build GRCh38
rs764994112 Chr.19: 44157296-44157296 on Build GRCh38
rs35570987 Chr.19: 44157285-44157286 on Build GRCh38
rs376282922 Chr.19: 44157267-44157267 on Build GRCh38
rs191045580 Chr.19: 44157170-44157170 on Build GRCh38
rs868428491 Chr.19: 44157190-44157190 on Build GRCh38
rs776221545 Chr.19: 44157245-44157245 on Build GRCh38
rs1050998717 Chr.19: 44157328-44157328 on Build GRCh38

View Full Product Details