Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 21076197-21076436 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF40
Entrez Gene ID
55701
Gene Name
Rho guanine nucleotide exchange factor 40
Gene Aliases
SOLO
UniGene
Hs.35125
Gene Chromosome Location
Chr.14: 21061276-21090248 on Build GRCh38
Transcripts
NM_001278529.1 NM_001278530.1 NM_018071.4 XM_005267844.2 XM_011536937.2 XM_011536938.2 XM_017021434.1 XM_017021435.1 XM_017021436.1 XM_017021437.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6030192 c.1767C>T p.S589S Chr.14: 21076387-21076387 on Build GRCh38
2029521 c.1757T>C p.L586P Chr.14: 21076377-21076377 on Build GRCh38
5493135 c.1783C>T p.R595C Chr.14: 21076403-21076403 on Build GRCh38
4381830 c.1766C>A p.S589Y Chr.14: 21076386-21076386 on Build GRCh38
954346 c.1798C>T p.L600L Chr.14: 21076418-21076418 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs114352616 Chr.14: 21076387-21076387 on Build GRCh38
rs761818082 Chr.14: 21076315-21076318 on Build GRCh38
rs762302190 Chr.14: 21076323-21076323 on Build GRCh38
rs148519122 Chr.14: 21076381-21076381 on Build GRCh38
rs765215059 Chr.14: 21076318-21076319 on Build GRCh38
rs780625866 Chr.14: 21076403-21076403 on Build GRCh38
rs565728745 Chr.14: 21076377-21076377 on Build GRCh38
rs762743669 Chr.14: 21076416-21076416 on Build GRCh38
rs1000226191 Chr.14: 21076328-21076328 on Build GRCh38
rs775822130 Chr.14: 21076330-21076330 on Build GRCh38
rs772592753 Chr.14: 21076327-21076327 on Build GRCh38
rs534855419 Chr.14: 21076388-21076388 on Build GRCh38
rs777103989 Chr.14: 21076414-21076414 on Build GRCh38
rs755946062 Chr.14: 21076375-21076375 on Build GRCh38
rs569320020 Chr.14: 21076282-21076282 on Build GRCh38
rs796578428 Chr.14: 21076318-21076319 on Build GRCh38
rs758425304 Chr.14: 21076404-21076404 on Build GRCh38
rs745982666 Chr.14: 21076393-21076393 on Build GRCh38
rs772975629 Chr.14: 21076350-21076350 on Build GRCh38
rs764308205 Chr.14: 21076345-21076345 on Build GRCh38
rs182818490 Chr.14: 21076356-21076356 on Build GRCh38
rs531132123 Chr.14: 21076238-21076238 on Build GRCh38
rs549332498 Chr.14: 21076239-21076239 on Build GRCh38
rs144677985 Chr.14: 21076364-21076364 on Build GRCh38
rs369724419 Chr.14: 21076360-21076360 on Build GRCh38
rs1052034498 Chr.14: 21076236-21076236 on Build GRCh38
rs200187872 Chr.14: 21076337-21076337 on Build GRCh38
rs746425111 Chr.14: 21076311-21076311 on Build GRCh38
rs762688751 Chr.14: 21076355-21076355 on Build GRCh38
rs986198382 Chr.14: 21076283-21076283 on Build GRCh38
rs767518133 Chr.14: 21076361-21076361 on Build GRCh38
rs1017553350 Chr.14: 21076258-21076258 on Build GRCh38
rs771980877 Chr.14: 21076394-21076394 on Build GRCh38

View Full Product Details