Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 34894949-34895190 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APIP
Entrez Gene ID
51074
Gene Name
APAF1 interacting protein
Gene Aliases
APIP2, CGI-29, CGI29, MMRP19, hAPIP
UniGene
Hs.447794
Gene Chromosome Location
Chr.11: 34882295-34916411 on Build GRCh38
Transcripts
NM_015957.3 XM_011520154.2 XM_017017875.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3752344 c.58G>A p.D20N Chr.11: 34895110-34895110 on Build GRCh38
3446984 c.58-1G>A p.? Chr.11: 34895111-34895111 on Build GRCh38
6461781 c.146G>T p.S49I Chr.11: 34895022-34895022 on Build GRCh38
5815611 c.196A>T p.S66C Chr.11: 34895023-34895023 on Build GRCh38
3716792 c.158+4A>C p.? Chr.11: 34895006-34895006 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs533209732 Chr.11: 34895093-34895093 on Build GRCh38
rs757244492 Chr.11: 34895058-34895058 on Build GRCh38
rs752124751 Chr.11: 34895028-34895028 on Build GRCh38
rs968938552 Chr.11: 34895071-34895071 on Build GRCh38
rs761893388 Chr.11: 34894993-34894993 on Build GRCh38
rs368305932 Chr.11: 34895154-34895154 on Build GRCh38
rs765529205 Chr.11: 34894994-34894994 on Build GRCh38
rs568309218 Chr.11: 34895155-34895155 on Build GRCh38
rs768096884 Chr.11: 34895146-34895146 on Build GRCh38
rs148171361 Chr.11: 34895057-34895057 on Build GRCh38
rs748974469 Chr.11: 34895065-34895065 on Build GRCh38
rs17850326 Chr.11: 34895100-34895100 on Build GRCh38
rs770919303 Chr.11: 34895084-34895084 on Build GRCh38
rs750689149 Chr.11: 34895009-34895009 on Build GRCh38
rs776534472 Chr.11: 34895105-34895105 on Build GRCh38
rs763298096 Chr.11: 34895122-34895122 on Build GRCh38
rs772161868 Chr.11: 34895102-34895102 on Build GRCh38
rs778044901 Chr.11: 34895056-34895056 on Build GRCh38
rs759987117 Chr.11: 34895145-34895145 on Build GRCh38
rs61734605 Chr.11: 34895110-34895110 on Build GRCh38
rs368297989 Chr.11: 34895015-34895015 on Build GRCh38
rs766856746 Chr.11: 34895020-34895020 on Build GRCh38
rs752271072 Chr.11: 34895049-34895049 on Build GRCh38
rs143796671 Chr.11: 34895038-34895038 on Build GRCh38
rs546676572 Chr.11: 34895109-34895109 on Build GRCh38
rs766354808 Chr.11: 34895005-34895005 on Build GRCh38
rs745854468 Chr.11: 34895086-34895086 on Build GRCh38
rs142818890 Chr.11: 34895127-34895127 on Build GRCh38
rs1030468887 Chr.11: 34894975-34894975 on Build GRCh38
rs773542191 Chr.11: 34895119-34895119 on Build GRCh38
rs148981873 Chr.11: 34895029-34895029 on Build GRCh38
rs752038800 Chr.11: 34895130-34895130 on Build GRCh38
rs1326941 Chr.11: 34895163-34895163 on Build GRCh38
rs770431311 Chr.11: 34895161-34895162 on Build GRCh38
rs760244671 Chr.11: 34895129-34895130 on Build GRCh38

View Full Product Details