Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 41246840-41247093 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ULK4
Entrez Gene ID
54986
Gene Name
unc-51 like kinase 4
Gene Aliases
FAM7C1, REC01035
UniGene
Hs.656192
Gene Chromosome Location
Chr.3: 41246599-41963020 on Build GRCh38
Transcripts
NM_001322501.1 NM_017886.3 NR_136342.1 XM_011533880.2 XM_017006711.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6257820 c.1264C>A p.L422I Chr.3: 41246949-41246949 on Build GRCh38
4647015 c.1273G>A p.V425I Chr.3: 41246940-41246940 on Build GRCh38
6450460 c.1271C>T p.A424V Chr.3: 41246942-41246942 on Build GRCh38
6257819 c.3808C>A p.L1270I Chr.3: 41246949-41246949 on Build GRCh38
4453982 c.1269A>G p.Q423Q Chr.3: 41246944-41246944 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs902415646 Chr.3: 41247052-41247052 on Build GRCh38
rs1011578798 Chr.3: 41246892-41246892 on Build GRCh38
rs756103924 Chr.3: 41246946-41246946 on Build GRCh38
rs761363601 Chr.3: 41246903-41246903 on Build GRCh38
rs748089332 Chr.3: 41246944-41246944 on Build GRCh38
rs781157144 Chr.3: 41246941-41246941 on Build GRCh38
rs937510760 Chr.3: 41246983-41246983 on Build GRCh38
rs376498555 Chr.3: 41246945-41246945 on Build GRCh38
rs143939899 Chr.3: 41247064-41247065 on Build GRCh38
rs764657225 Chr.3: 41246884-41246884 on Build GRCh38
rs202237555 Chr.3: 41246981-41246981 on Build GRCh38
rs754342213 Chr.3: 41246894-41246894 on Build GRCh38
rs373420715 Chr.3: 41246910-41246910 on Build GRCh38
rs766128364 Chr.3: 41247003-41247003 on Build GRCh38
rs751389280 Chr.3: 41246935-41246935 on Build GRCh38
rs745956482 Chr.3: 41246970-41246970 on Build GRCh38
rs772266649 Chr.3: 41246974-41246974 on Build GRCh38
rs761083190 Chr.3: 41246883-41246883 on Build GRCh38
rs893227743 Chr.3: 41247018-41247018 on Build GRCh38
rs878878579 Chr.3: 41247047-41247047 on Build GRCh38
rs775772003 Chr.3: 41246881-41246881 on Build GRCh38
rs755944153 Chr.3: 41247033-41247033 on Build GRCh38
rs12488691 Chr.3: 41246978-41246978 on Build GRCh38
rs529574284 Chr.3: 41247019-41247019 on Build GRCh38
rs6769117 Chr.3: 41246975-41246975 on Build GRCh38
rs754783012 Chr.3: 41247017-41247017 on Build GRCh38
rs777075861 Chr.3: 41246979-41246979 on Build GRCh38
rs1028704623 Chr.3: 41246933-41246933 on Build GRCh38
rs369238979 Chr.3: 41246900-41246900 on Build GRCh38
rs752232083 Chr.3: 41246906-41246911 on Build GRCh38
rs752435686 Chr.3: 41246952-41246952 on Build GRCh38
rs754890627 Chr.3: 41246940-41246940 on Build GRCh38
rs533518431 Chr.3: 41246932-41246932 on Build GRCh38
rs141687204 Chr.3: 41247070-41247070 on Build GRCh38
rs371027647 Chr.3: 41247063-41247064 on Build GRCh38
rs879188320 Chr.3: 41247064-41247064 on Build GRCh38
rs529343221 Chr.3: 41246906-41246906 on Build GRCh38
rs375325027 Chr.3: 41246921-41246921 on Build GRCh38
rs766362471 Chr.3: 41246904-41246904 on Build GRCh38
rs751299549 Chr.3: 41247013-41247013 on Build GRCh38
rs371158713 Chr.3: 41246982-41246982 on Build GRCh38
rs762612674 Chr.3: 41246991-41246991 on Build GRCh38
rs373109226 Chr.3: 41247024-41247024 on Build GRCh38
rs189499822 Chr.3: 41247046-41247046 on Build GRCh38
rs372547665 Chr.3: 41246976-41246976 on Build GRCh38

View Full Product Details