Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 22180588-22180837 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF676
Entrez Gene ID
163223
Gene Name
zinc finger protein 676
Gene Aliases
-
UniGene
Hs.55452
Gene Chromosome Location
Chr.19: 22179101-22196951 on Build GRCh38
Transcripts
NM_001001411.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3756559 c.1106G>A p.G369E Chr.19: 22180611-22180611 on Build GRCh38
1271691 c.1091C>A p.P364H Chr.19: 22180626-22180626 on Build GRCh38
4170997 c.1128G>C p.K376N Chr.19: 22180589-22180589 on Build GRCh38
3773851 c.888C>T p.L296L Chr.19: 22180829-22180829 on Build GRCh38
338586 c.1000G>A p.E334K Chr.19: 22180717-22180717 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs548029407 Chr.19: 22180718-22180718 on Build GRCh38
rs77338425 Chr.19: 22180688-22180688 on Build GRCh38
rs764780050 Chr.19: 22180615-22180615 on Build GRCh38
rs778184207 Chr.19: 22180725-22180725 on Build GRCh38
rs78757874 Chr.19: 22180658-22180658 on Build GRCh38
rs758728714 Chr.19: 22180719-22180719 on Build GRCh38
rs767350295 Chr.19: 22180804-22180804 on Build GRCh38
rs758119822 Chr.19: 22180626-22180626 on Build GRCh38
rs750271616 Chr.19: 22180806-22180806 on Build GRCh38
rs777247559 Chr.19: 22180668-22180668 on Build GRCh38
rs752505322 Chr.19: 22180808-22180808 on Build GRCh38
rs751324293 Chr.19: 22180703-22180703 on Build GRCh38
rs779066312 Chr.19: 22180715-22180715 on Build GRCh38
rs766099109 Chr.19: 22180775-22180775 on Build GRCh38
rs78050927 Chr.19: 22180706-22180706 on Build GRCh38
rs781229304 Chr.19: 22180645-22180645 on Build GRCh38
rs200452805 Chr.19: 22180646-22180646 on Build GRCh38
rs566378841 Chr.19: 22180736-22180736 on Build GRCh38
rs762487915 Chr.19: 22180753-22180753 on Build GRCh38
rs564215111 Chr.19: 22180648-22180648 on Build GRCh38
rs75690456 Chr.19: 22180611-22180611 on Build GRCh38
rs764804869 Chr.19: 22180674-22180674 on Build GRCh38
rs762615204 Chr.19: 22180690-22180690 on Build GRCh38
rs184803507 Chr.19: 22180643-22180643 on Build GRCh38
rs373247831 Chr.19: 22180807-22180807 on Build GRCh38
rs754948281 Chr.19: 22180712-22180712 on Build GRCh38
rs550042173 Chr.19: 22180667-22180667 on Build GRCh38
rs775011020 Chr.19: 22180748-22180748 on Build GRCh38
rs767344565 Chr.19: 22180710-22180710 on Build GRCh38
rs769133931 Chr.19: 22180737-22180737 on Build GRCh38
rs752457199 Chr.19: 22180618-22180618 on Build GRCh38
rs267605390 Chr.19: 22180642-22180642 on Build GRCh38
rs765193548 Chr.19: 22180809-22180809 on Build GRCh38
rs751448887 Chr.19: 22180636-22180636 on Build GRCh38
rs748334900 Chr.19: 22180716-22180716 on Build GRCh38
rs773737407 Chr.19: 22180665-22180665 on Build GRCh38
rs373787022 Chr.19: 22180664-22180664 on Build GRCh38
rs777555826 Chr.19: 22180627-22180627 on Build GRCh38
rs201685218 Chr.19: 22180675-22180675 on Build GRCh38
rs760383294 Chr.19: 22180723-22180723 on Build GRCh38
rs80146743 Chr.19: 22180787-22180787 on Build GRCh38
rs568421167 Chr.19: 22180670-22180670 on Build GRCh38
rs201622264 Chr.19: 22180808-22180808 on Build GRCh38
rs369672319 Chr.19: 22180735-22180735 on Build GRCh38
rs747360114 Chr.19: 22180727-22180727 on Build GRCh38
rs759129494 Chr.19: 22180612-22180612 on Build GRCh38
rs774048823 Chr.19: 22180803-22180803 on Build GRCh38
rs778998724 Chr.19: 22180655-22180655 on Build GRCh38
rs771388999 Chr.19: 22180734-22180734 on Build GRCh38
rs78855594 Chr.19: 22180691-22180691 on Build GRCh38

View Full Product Details