Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 38356409-38356676 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CATSPERG
Entrez Gene ID
57828
Gene Name
cation channel sperm associated auxiliary subunit gamma
Gene Aliases
C19orf15
UniGene
Hs.324335
Gene Chromosome Location
Chr.19: 38335563-38370949 on Build GRCh38
Transcripts
NM_021185.4 XM_005259114.4 XM_005259115.4 XM_005259116.2 XM_006723308.3 XM_006723310.2 XM_006723311.2 XM_011527174.2 XM_011527175.2 XM_011527176.2 XM_011527177.2 XM_011527178.2 XM_011527179.1 XM_011527182.2 XM_017027053.1 XM_017027054.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6620822 c.81G>A p.L27L Chr.19: 38356509-38356509 on Build GRCh38
1393340 c.102C>T p.C34C Chr.19: 38356530-38356530 on Build GRCh38
711653 c.103G>T p.E35* Chr.19: 38356531-38356531 on Build GRCh38
5775841 c.1165A>T p.R389W Chr.19: 38356513-38356513 on Build GRCh38
3533317 c.83C>T p.P28L Chr.19: 38356511-38356511 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373175131 Chr.19: 38356468-38356468 on Build GRCh38
rs776412128 Chr.19: 38356494-38356494 on Build GRCh38
rs758454852 Chr.19: 38356582-38356582 on Build GRCh38
rs370849392 Chr.19: 38356547-38356547 on Build GRCh38
rs748237417 Chr.19: 38356479-38356479 on Build GRCh38
rs981559559 Chr.19: 38356641-38356641 on Build GRCh38
rs749456391 Chr.19: 38356437-38356437 on Build GRCh38
rs964771156 Chr.19: 38356539-38356539 on Build GRCh38
rs184862901 Chr.19: 38356568-38356568 on Build GRCh38
rs762854465 Chr.19: 38356453-38356453 on Build GRCh38
rs768920618 Chr.19: 38356454-38356454 on Build GRCh38
rs373128819 Chr.19: 38356530-38356530 on Build GRCh38
rs368740414 Chr.19: 38356469-38356469 on Build GRCh38
rs545356454 Chr.19: 38356630-38356630 on Build GRCh38
rs755348839 Chr.19: 38356569-38356569 on Build GRCh38
rs763548211 Chr.19: 38356472-38356472 on Build GRCh38
rs370127051 Chr.19: 38356570-38356570 on Build GRCh38
rs1011185566 Chr.19: 38356462-38356462 on Build GRCh38
rs771024924 Chr.19: 38356518-38356521 on Build GRCh38
rs1028348821 Chr.19: 38356548-38356548 on Build GRCh38
rs142066780 Chr.19: 38356557-38356557 on Build GRCh38
rs879103683 Chr.19: 38356616-38356616 on Build GRCh38
rs747239141 Chr.19: 38356447-38356448 on Build GRCh38
rs957875529 Chr.19: 38356643-38356643 on Build GRCh38
rs770653802 Chr.19: 38356492-38356492 on Build GRCh38
rs777842326 Chr.19: 38356586-38356586 on Build GRCh38
rs748349005 Chr.19: 38356576-38356576 on Build GRCh38
rs750906220 Chr.19: 38356556-38356556 on Build GRCh38
rs767061763 Chr.19: 38356558-38356558 on Build GRCh38
rs773092595 Chr.19: 38356486-38356486 on Build GRCh38
rs141054803 Chr.19: 38356516-38356516 on Build GRCh38
rs772083470 Chr.19: 38356481-38356481 on Build GRCh38
rs746970093 Chr.19: 38356490-38356490 on Build GRCh38
rs764925813 Chr.19: 38356521-38356521 on Build GRCh38
rs763724054 Chr.19: 38356550-38356550 on Build GRCh38
rs894851378 Chr.19: 38356526-38356526 on Build GRCh38
rs996423100 Chr.19: 38356545-38356545 on Build GRCh38
rs146338932 Chr.19: 38356611-38356611 on Build GRCh38

View Full Product Details