Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 92353689-92353934 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAT3
Entrez Gene ID
120114
Gene Name
FAT atypical cadherin 3
Gene Aliases
CDHF15, CDHR10, hFat3
UniGene
Hs.98523
Gene Chromosome Location
Chr.11: 92224636-92896473 on Build GRCh38
Transcripts
NM_001008781.2 XM_017017178.1 XM_017017179.1 XM_017017180.1 XM_017017181.1 XM_017017182.1 XM_017017183.1 XM_017017184.1 XM_017017186.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6135264 c.1669C>A p.R557S Chr.11: 92353781-92353781 on Build GRCh38
2223243 c.1649C>G p.S550C Chr.11: 92353761-92353761 on Build GRCh38
5106047 c.1593T>C p.T531T Chr.11: 92353705-92353705 on Build GRCh38
6205676 c.1763C>A p.S588* Chr.11: 92353875-92353875 on Build GRCh38
3870405 c.1754G>A p.G585E Chr.11: 92353866-92353866 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750771501 Chr.11: 92353867-92353867 on Build GRCh38
rs552221485 Chr.11: 92353834-92353834 on Build GRCh38
rs768045526 Chr.11: 92353863-92353863 on Build GRCh38
rs745503979 Chr.11: 92353906-92353906 on Build GRCh38
rs764641369 Chr.11: 92353856-92353856 on Build GRCh38
rs753989720 Chr.11: 92353876-92353876 on Build GRCh38
rs75759020 Chr.11: 92353860-92353860 on Build GRCh38
rs763402792 Chr.11: 92353748-92353748 on Build GRCh38
rs913907904 Chr.11: 92353758-92353758 on Build GRCh38
rs770533228 Chr.11: 92353829-92353829 on Build GRCh38
rs201479806 Chr.11: 92353870-92353870 on Build GRCh38
rs780305849 Chr.11: 92353782-92353782 on Build GRCh38
rs372561498 Chr.11: 92353849-92353849 on Build GRCh38
rs767992417 Chr.11: 92353777-92353777 on Build GRCh38
rs776156851 Chr.11: 92353735-92353735 on Build GRCh38
rs368210310 Chr.11: 92353900-92353900 on Build GRCh38
rs748165817 Chr.11: 92353892-92353892 on Build GRCh38
rs72970548 Chr.11: 92353828-92353828 on Build GRCh38
rs868062627 Chr.11: 92353740-92353740 on Build GRCh38
rs267603240 Chr.11: 92353812-92353812 on Build GRCh38
rs757698325 Chr.11: 92353759-92353759 on Build GRCh38
rs771837329 Chr.11: 92353721-92353721 on Build GRCh38
rs370173528 Chr.11: 92353790-92353790 on Build GRCh38
rs777574389 Chr.11: 92353899-92353899 on Build GRCh38
rs758281021 Chr.11: 92353893-92353893 on Build GRCh38
rs776403254 Chr.11: 92353905-92353905 on Build GRCh38
rs778826850 Chr.11: 92353886-92353886 on Build GRCh38
rs751884222 Chr.11: 92353749-92353749 on Build GRCh38
rs748030116 Chr.11: 92353802-92353802 on Build GRCh38
rs754881712 Chr.11: 92353797-92353797 on Build GRCh38
rs905724342 Chr.11: 92353751-92353751 on Build GRCh38
rs765921423 Chr.11: 92353734-92353734 on Build GRCh38
rs749319415 Chr.11: 92353785-92353785 on Build GRCh38
rs755149520 Chr.11: 92353885-92353885 on Build GRCh38
rs766563317 Chr.11: 92353871-92353871 on Build GRCh38
rs769390532 Chr.11: 92353907-92353907 on Build GRCh38
rs367795074 Chr.11: 92353733-92353733 on Build GRCh38
rs975021676 Chr.11: 92353887-92353887 on Build GRCh38
rs756385214 Chr.11: 92353781-92353781 on Build GRCh38
rs376278280 Chr.11: 92353800-92353800 on Build GRCh38
rs770586274 Chr.11: 92353904-92353904 on Build GRCh38

View Full Product Details