Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 163513583-163513856 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAT2B
Entrez Gene ID
27430
Gene Name
methionine adenosyltransferase 2B
Gene Aliases
MAT-II, MATIIbeta, Nbla02999, SDR23E1, TGR
UniGene
Hs.54642
Gene Chromosome Location
Chr.5: 163503064-163519353 on Build GRCh38
Transcripts
NM_013283.4 NM_182796.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5737413 c.320G>T p.G107V Chr.5: 163513649-163513649 on Build GRCh38
6031236 c.330A>G p.A110A Chr.5: 163513659-163513659 on Build GRCh38
6031237 c.363A>G p.A121A Chr.5: 163513659-163513659 on Build GRCh38
3722484 c.311A>T p.N104I Chr.5: 163513607-163513607 on Build GRCh38
98299 c.278A>T p.N93I Chr.5: 163513607-163513607 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766779756 Chr.5: 163513675-163513675 on Build GRCh38
rs895134805 Chr.5: 163513804-163513804 on Build GRCh38
rs760659097 Chr.5: 163513617-163513617 on Build GRCh38
rs776924215 Chr.5: 163513799-163513799 on Build GRCh38
rs752246412 Chr.5: 163513700-163513700 on Build GRCh38
rs772273855 Chr.5: 163513801-163513803 on Build GRCh38
rs775707428 Chr.5: 163513612-163513612 on Build GRCh38
rs116612015 Chr.5: 163513711-163513711 on Build GRCh38
rs7729611 Chr.5: 163513739-163513739 on Build GRCh38
rs946157791 Chr.5: 163513717-163513717 on Build GRCh38
rs771051976 Chr.5: 163513719-163513719 on Build GRCh38
rs926146971 Chr.5: 163513735-163513735 on Build GRCh38
rs748353115 Chr.5: 163513797-163513799 on Build GRCh38
rs113852542 Chr.5: 163513793-163513793 on Build GRCh38
rs113548741 Chr.5: 163513629-163513629 on Build GRCh38
rs748301386 Chr.5: 163513805-163513805 on Build GRCh38
rs866869648 Chr.5: 163513619-163513619 on Build GRCh38
rs781428598 Chr.5: 163513625-163513625 on Build GRCh38
rs141130487 Chr.5: 163513659-163513659 on Build GRCh38
rs773728952 Chr.5: 163513809-163513810 on Build GRCh38
rs747154409 Chr.5: 163513795-163513795 on Build GRCh38
rs755909996 Chr.5: 163513709-163513709 on Build GRCh38
rs754411771 Chr.5: 163513686-163513686 on Build GRCh38
rs765526423 Chr.5: 163513640-163513640 on Build GRCh38
rs964945 Chr.5: 163513733-163513733 on Build GRCh38
rs768569660 Chr.5: 163513797-163513797 on Build GRCh38
rs778000635 Chr.5: 163513812-163513812 on Build GRCh38
rs931383079 Chr.5: 163513747-163513750 on Build GRCh38
rs768973562 Chr.5: 163513623-163513623 on Build GRCh38
rs879221826 Chr.5: 163513743-163513743 on Build GRCh38
rs191980671 Chr.5: 163513674-163513674 on Build GRCh38
rs776399886 Chr.5: 163513831-163513831 on Build GRCh38
rs370041394 Chr.5: 163513683-163513683 on Build GRCh38
rs771419964 Chr.5: 163513830-163513831 on Build GRCh38
rs763277709 Chr.5: 163513830-163513830 on Build GRCh38
rs749092287 Chr.5: 163513718-163513718 on Build GRCh38
rs992759230 Chr.5: 163513679-163513679 on Build GRCh38
rs761182503 Chr.5: 163513826-163513828 on Build GRCh38
rs780932292 Chr.5: 163513695-163513695 on Build GRCh38
rs773622292 Chr.5: 163513818-163513818 on Build GRCh38

View Full Product Details