Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49476414-49476661 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLT3LG
Entrez Gene ID
2323
Gene Name
fms related tyrosine kinase 3 ligand
Gene Aliases
FL, FLT3L
UniGene
Hs.428
Gene Chromosome Location
Chr.19: 49474172-49487038 on Build GRCh38
Transcripts
NM_001204502.1 NM_001204503.1 NM_001278637.1 NM_001278638.1 NM_001459.3 XM_005258680.4 XM_005258681.4 XM_005258682.4 XM_005258683.4 XM_006723116.3 XM_011526675.2 XM_011526676.2 XM_011526677.2 XM_011526678.2 XM_011526680.2 XM_011526682.2 XM_017026532.1 XM_017026533.1 XM_017026534.1 XM_017026535.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4127784 c.203A>C p.E68A Chr.19: 49476427-49476427 on Build GRCh38
6004295 c.256C>T p.L86F Chr.19: 49476480-49476480 on Build GRCh38
6506117 c.225G>A p.R75R Chr.19: 49476449-49476449 on Build GRCh38
4940547 c.242G>A p.R81H Chr.19: 49476466-49476466 on Build GRCh38
1252586 c.211G>T p.G71W Chr.19: 49476435-49476435 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752160204 Chr.19: 49476522-49476522 on Build GRCh38
rs912230033 Chr.19: 49476439-49476439 on Build GRCh38
rs780247349 Chr.19: 49476604-49476604 on Build GRCh38
rs745514077 Chr.19: 49476583-49476583 on Build GRCh38
rs894541425 Chr.19: 49476630-49476630 on Build GRCh38
rs776638815 Chr.19: 49476434-49476434 on Build GRCh38
rs939916516 Chr.19: 49476477-49476477 on Build GRCh38
rs751911650 Chr.19: 49476481-49476481 on Build GRCh38
rs765687695 Chr.19: 49476450-49476450 on Build GRCh38
rs745652407 Chr.19: 49476609-49476609 on Build GRCh38
rs753850158 Chr.19: 49476525-49476525 on Build GRCh38
rs766783205 Chr.19: 49476478-49476478 on Build GRCh38
rs79058858 Chr.19: 49476532-49476532 on Build GRCh38
rs917193040 Chr.19: 49476491-49476491 on Build GRCh38
rs761882863 Chr.19: 49476448-49476448 on Build GRCh38
rs866630139 Chr.19: 49476455-49476455 on Build GRCh38
rs3826717 Chr.19: 49476636-49476636 on Build GRCh38
rs368893926 Chr.19: 49476515-49476515 on Build GRCh38
rs572235634 Chr.19: 49476602-49476602 on Build GRCh38
rs200833992 Chr.19: 49476590-49476590 on Build GRCh38
rs773732267 Chr.19: 49476473-49476473 on Build GRCh38
rs769003253 Chr.19: 49476616-49476616 on Build GRCh38
rs143173973 Chr.19: 49476492-49476492 on Build GRCh38
rs748209726 Chr.19: 49476617-49476617 on Build GRCh38
rs758534099 Chr.19: 49476533-49476533 on Build GRCh38
rs754695522 Chr.19: 49476487-49476487 on Build GRCh38
rs139793754 Chr.19: 49476475-49476475 on Build GRCh38
rs755598105 Chr.19: 49476523-49476523 on Build GRCh38
rs777599465 Chr.19: 49476524-49476524 on Build GRCh38
rs868574879 Chr.19: 49476435-49476435 on Build GRCh38
rs931124316 Chr.19: 49476574-49476574 on Build GRCh38
rs1055623560 Chr.19: 49476488-49476488 on Build GRCh38
rs763423681 Chr.19: 49476476-49476476 on Build GRCh38

View Full Product Details