Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32247315-32247564 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM167B
Entrez Gene ID
84734
Gene Name
family with sequence similarity 167 member B
Gene Aliases
C1orf90
UniGene
Hs.18449
Gene Chromosome Location
Chr.1: 32247217-32248860 on Build GRCh38
Transcripts
NM_032648.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
426095 c.77C>T p.S26F Chr.1: 32247498-32247498 on Build GRCh38
6331233 c.69C>A p.S23R Chr.1: 32247490-32247490 on Build GRCh38
70699 c.61G>T p.G21W Chr.1: 32247482-32247482 on Build GRCh38
1667348 c.79G>A p.V27M Chr.1: 32247500-32247500 on Build GRCh38
7199100 c.27G>C p.Q9H Chr.1: 32247448-32247448 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757563076 Chr.1: 32247420-32247420 on Build GRCh38
rs371605638 Chr.1: 32247532-32247532 on Build GRCh38
rs774385135 Chr.1: 32247497-32247497 on Build GRCh38
rs780497345 Chr.1: 32247454-32247454 on Build GRCh38
rs772865076 Chr.1: 32247478-32247478 on Build GRCh38
rs769409836 Chr.1: 32247373-32247373 on Build GRCh38
rs549437991 Chr.1: 32247540-32247540 on Build GRCh38
rs368102614 Chr.1: 32247467-32247467 on Build GRCh38
rs371952094 Chr.1: 32247396-32247396 on Build GRCh38
rs773126596 Chr.1: 32247379-32247379 on Build GRCh38
rs758799260 Chr.1: 32247447-32247447 on Build GRCh38
rs770666613 Chr.1: 32247489-32247489 on Build GRCh38
rs752797339 Chr.1: 32247399-32247399 on Build GRCh38
rs760799856 Chr.1: 32247403-32247403 on Build GRCh38
rs764280624 Chr.1: 32247410-32247410 on Build GRCh38
rs777293745 Chr.1: 32247376-32247376 on Build GRCh38
rs758739525 Chr.1: 32247543-32247543 on Build GRCh38
rs745833405 Chr.1: 32247499-32247500 on Build GRCh38
rs761470712 Chr.1: 32247372-32247372 on Build GRCh38
rs754044304 Chr.1: 32247415-32247415 on Build GRCh38
rs764262284 Chr.1: 32247529-32247529 on Build GRCh38
rs201866460 Chr.1: 32247466-32247466 on Build GRCh38
rs1020406675 Chr.1: 32247365-32247365 on Build GRCh38
rs765535479 Chr.1: 32247539-32247539 on Build GRCh38
rs961298997 Chr.1: 32247534-32247534 on Build GRCh38
rs200643978 Chr.1: 32247535-32247535 on Build GRCh38
rs375921844 Chr.1: 32247536-32247536 on Build GRCh38
rs747534830 Chr.1: 32247465-32247465 on Build GRCh38
rs547267308 Chr.1: 32247457-32247457 on Build GRCh38
rs762790238 Chr.1: 32247383-32247383 on Build GRCh38
rs749152074 Chr.1: 32247483-32247483 on Build GRCh38
rs769544178 Chr.1: 32247475-32247475 on Build GRCh38
rs759548407 Chr.1: 32247508-32247508 on Build GRCh38
rs201497447 Chr.1: 32247472-32247472 on Build GRCh38
rs765501225 Chr.1: 32247426-32247426 on Build GRCh38
rs41264003 Chr.1: 32247432-32247432 on Build GRCh38

View Full Product Details