Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 90904147-90904406 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAN2A2
Entrez Gene ID
4122
Gene Name
mannosidase alpha class 2A member 2
Gene Aliases
MANA2X
UniGene
Hs.116459
Gene Chromosome Location
Chr.15: 90902218-90922585 on Build GRCh38
Transcripts
NM_001320977.1 NM_006122.3 NR_135502.1 NR_135503.1 NR_135504.1 XM_005254910.3 XM_017022184.1 XM_017022185.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6312740 c.29G>T p.C10F Chr.15: 90904236-90904236 on Build GRCh38
1180297 c.111G>C p.Q37H Chr.15: 90904318-90904318 on Build GRCh38
2155635 c.53C>G p.A18G Chr.15: 90904260-90904260 on Build GRCh38
2155636 c.52G>A p.A18T Chr.15: 90904259-90904259 on Build GRCh38
6706641 c.97C>T p.P33S Chr.15: 90904304-90904304 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs987225260 Chr.15: 90904315-90904315 on Build GRCh38
rs771074645 Chr.15: 90904333-90904333 on Build GRCh38
rs2677743 Chr.15: 90904279-90904279 on Build GRCh38
rs778964870 Chr.15: 90904313-90904313 on Build GRCh38
rs376674936 Chr.15: 90904311-90904311 on Build GRCh38
rs764470576 Chr.15: 90904353-90904353 on Build GRCh38
rs756805277 Chr.15: 90904185-90904185 on Build GRCh38
rs369935789 Chr.15: 90904180-90904180 on Build GRCh38
rs111645863 Chr.15: 90904195-90904195 on Build GRCh38
rs1017126552 Chr.15: 90904227-90904227 on Build GRCh38
rs780789542 Chr.15: 90904194-90904194 on Build GRCh38
rs942665615 Chr.15: 90904365-90904365 on Build GRCh38
rs746027112 Chr.15: 90904338-90904338 on Build GRCh38
rs201522267 Chr.15: 90904346-90904346 on Build GRCh38
rs934052204 Chr.15: 90904301-90904301 on Build GRCh38
rs748128678 Chr.15: 90904317-90904317 on Build GRCh38
rs747049723 Chr.15: 90904182-90904182 on Build GRCh38
rs970300113 Chr.15: 90904220-90904220 on Build GRCh38
rs766948945 Chr.15: 90904376-90904376 on Build GRCh38
rs755892981 Chr.15: 90904199-90904199 on Build GRCh38
rs916840965 Chr.15: 90904281-90904281 on Build GRCh38
rs373992965 Chr.15: 90904238-90904238 on Build GRCh38
rs763432859 Chr.15: 90904277-90904277 on Build GRCh38
rs775713748 Chr.15: 90904270-90904270 on Build GRCh38
rs779729644 Chr.15: 90904200-90904200 on Build GRCh38
rs980292769 Chr.15: 90904223-90904223 on Build GRCh38
rs925450524 Chr.15: 90904201-90904201 on Build GRCh38
rs113662660 Chr.15: 90904336-90904336 on Build GRCh38
rs779827251 Chr.15: 90904299-90904299 on Build GRCh38
rs750038021 Chr.15: 90904292-90904292 on Build GRCh38
rs759690298 Chr.15: 90904253-90904253 on Build GRCh38
rs774788334 Chr.15: 90904360-90904360 on Build GRCh38
rs776954641 Chr.15: 90904244-90904244 on Build GRCh38
rs764652094 Chr.15: 90904179-90904179 on Build GRCh38
rs762324234 Chr.15: 90904374-90904374 on Build GRCh38
rs745548359 Chr.15: 90904198-90904198 on Build GRCh38
rs749900448 Chr.15: 90904386-90904386 on Build GRCh38
rs752096187 Chr.15: 90904288-90904288 on Build GRCh38
rs774489487 Chr.15: 90904217-90904217 on Build GRCh38
rs768905346 Chr.15: 90904295-90904295 on Build GRCh38
rs756713868 Chr.15: 90904289-90904289 on Build GRCh38
rs776852590 Chr.15: 90904337-90904337 on Build GRCh38
rs1043688868 Chr.15: 90904382-90904382 on Build GRCh38
rs199595126 Chr.15: 90904312-90904312 on Build GRCh38
rs768881302 Chr.15: 90904231-90904232 on Build GRCh38
rs749023505 Chr.15: 90904310-90904310 on Build GRCh38
rs763191962 Chr.15: 90904351-90904351 on Build GRCh38
rs1046795056 Chr.15: 90904335-90904335 on Build GRCh38
rs978482224 Chr.15: 90904186-90904186 on Build GRCh38
rs755729151 Chr.15: 90904293-90904293 on Build GRCh38
rs560640558 Chr.15: 90904269-90904269 on Build GRCh38
rs749186537 Chr.15: 90904203-90904203 on Build GRCh38
rs768477616 Chr.15: 90904204-90904204 on Build GRCh38
rs542275390 Chr.15: 90904224-90904224 on Build GRCh38

View Full Product Details