Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18309647-18309886 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LSM4
Entrez Gene ID
25804
Gene Name
LSM4 homolog, U6 small nuclear RNA and mRNA degradation associated
Gene Aliases
GRP, YER112W
UniGene
Hs.515255
Gene Chromosome Location
Chr.19: 18306230-18323191 on Build GRCh38
Transcripts
NM_001252129.1 NM_012321.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4075663 c.264C>T p.R88R Chr.19: 18309742-18309742 on Build GRCh38
6246999 c.204C>A p.Y68* Chr.19: 18309802-18309802 on Build GRCh38
3530792 c.312G>A p.M104I Chr.19: 18309694-18309694 on Build GRCh38
4131471 c.180C>A p.Y60* Chr.19: 18309826-18309826 on Build GRCh38
6989139 c.308G>A p.G103D Chr.19: 18309698-18309698 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770100426 Chr.19: 18309718-18309720 on Build GRCh38
rs773289479 Chr.19: 18309737-18309737 on Build GRCh38
rs367937303 Chr.19: 18309769-18309769 on Build GRCh38
rs371805780 Chr.19: 18309796-18309796 on Build GRCh38
rs138296949 Chr.19: 18309778-18309778 on Build GRCh38
rs750013273 Chr.19: 18309680-18309680 on Build GRCh38
rs765934857 Chr.19: 18309742-18309742 on Build GRCh38
rs780227379 Chr.19: 18309865-18309865 on Build GRCh38
rs778056029 Chr.19: 18309767-18309767 on Build GRCh38
rs765738686 Chr.19: 18309688-18309688 on Build GRCh38
rs45578432 Chr.19: 18309821-18309821 on Build GRCh38
rs761687592 Chr.19: 18309735-18309735 on Build GRCh38
rs374309423 Chr.19: 18309809-18309809 on Build GRCh38
rs772004530 Chr.19: 18309791-18309791 on Build GRCh38
rs756153479 Chr.19: 18309859-18309859 on Build GRCh38
rs749456274 Chr.19: 18309790-18309790 on Build GRCh38
rs755682644 Chr.19: 18309686-18309686 on Build GRCh38
rs748477177 Chr.19: 18309694-18309694 on Build GRCh38
rs781537682 Chr.19: 18309842-18309842 on Build GRCh38
rs150898099 Chr.19: 18309743-18309743 on Build GRCh38
rs780420380 Chr.19: 18309789-18309789 on Build GRCh38
rs746224061 Chr.19: 18309710-18309711 on Build GRCh38
rs921443722 Chr.19: 18309827-18309827 on Build GRCh38
rs762546802 Chr.19: 18309828-18309828 on Build GRCh38
rs112426584 Chr.19: 18309863-18309863 on Build GRCh38
rs1012140285 Chr.19: 18309667-18309667 on Build GRCh38
rs758754967 Chr.19: 18309757-18309757 on Build GRCh38
rs757835207 Chr.19: 18309841-18309841 on Build GRCh38
rs148877950 Chr.19: 18309835-18309835 on Build GRCh38
rs964265072 Chr.19: 18309820-18309820 on Build GRCh38
rs761883527 Chr.19: 18309666-18309666 on Build GRCh38
rs750584733 Chr.19: 18309843-18309843 on Build GRCh38
rs769880716 Chr.19: 18309727-18309727 on Build GRCh38
rs148380214 Chr.19: 18309691-18309691 on Build GRCh38
rs754699075 Chr.19: 18309744-18309744 on Build GRCh38
rs765271867 Chr.19: 18309668-18309668 on Build GRCh38
rs540209324 Chr.19: 18309784-18309784 on Build GRCh38
rs767630097 Chr.19: 18309736-18309736 on Build GRCh38
rs141593327 Chr.19: 18309700-18309700 on Build GRCh38
rs867608609 Chr.19: 18309684-18309684 on Build GRCh38
rs764771715 Chr.19: 18309751-18309751 on Build GRCh38
rs774336443 Chr.19: 18309732-18309732 on Build GRCh38
rs1041158907 Chr.19: 18309819-18309819 on Build GRCh38
rs766768563 Chr.19: 18309787-18309787 on Build GRCh38
rs764476513 Chr.19: 18309834-18309834 on Build GRCh38
rs746136304 Chr.19: 18309709-18309709 on Build GRCh38
rs757668962 Chr.19: 18309774-18309774 on Build GRCh38
rs201611097 Chr.19: 18309866-18309866 on Build GRCh38
rs776526554 Chr.19: 18309702-18309702 on Build GRCh38
rs200432875 Chr.19: 18309802-18309802 on Build GRCh38
rs367855479 Chr.19: 18309822-18309822 on Build GRCh38
rs753540459 Chr.19: 18309690-18309690 on Build GRCh38
rs760630076 Chr.19: 18309741-18309741 on Build GRCh38
rs2232970 Chr.19: 18309699-18309699 on Build GRCh38
rs566635305 Chr.19: 18309760-18309760 on Build GRCh38
rs760697591 Chr.19: 18309797-18309797 on Build GRCh38
rs771059611 Chr.19: 18309701-18309701 on Build GRCh38

View Full Product Details