Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45590634-45590895 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR4
Entrez Gene ID
2828
Gene Name
G protein-coupled receptor 4
Gene Aliases
-
UniGene
Hs.17170
Gene Chromosome Location
Chr.19: 45589764-45602208 on Build GRCh38
Transcripts
NM_005282.2 XM_017026607.1 XM_017026608.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5856169 c.1019G>A p.G340D Chr.19: 45590848-45590848 on Build GRCh38
7250909 c.1054C>G p.Q352E Chr.19: 45590813-45590813 on Build GRCh38
6674187 c.998G>A p.S333N Chr.19: 45590869-45590869 on Build GRCh38
6316090 c.1054C>A p.Q352K Chr.19: 45590813-45590813 on Build GRCh38
128094 c.987C>T p.S329S Chr.19: 45590880-45590880 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145450390 Chr.19: 45590791-45590791 on Build GRCh38
rs759269013 Chr.19: 45590858-45590858 on Build GRCh38
rs917571690 Chr.19: 45590867-45590867 on Build GRCh38
rs371485406 Chr.19: 45590745-45590745 on Build GRCh38
rs201822609 Chr.19: 45590757-45590757 on Build GRCh38
rs147607317 Chr.19: 45590805-45590805 on Build GRCh38
rs150834740 Chr.19: 45590781-45590781 on Build GRCh38
rs369757213 Chr.19: 45590838-45590838 on Build GRCh38
rs886403167 Chr.19: 45590671-45590671 on Build GRCh38
rs374422009 Chr.19: 45590753-45590753 on Build GRCh38
rs139283755 Chr.19: 45590790-45590790 on Build GRCh38
rs943571939 Chr.19: 45590861-45590861 on Build GRCh38
rs774645776 Chr.19: 45590729-45590729 on Build GRCh38
rs761739112 Chr.19: 45590833-45590833 on Build GRCh38
rs551669816 Chr.19: 45590830-45590830 on Build GRCh38
rs776383785 Chr.19: 45590738-45590738 on Build GRCh38
rs751145496 Chr.19: 45590855-45590855 on Build GRCh38
rs528955502 Chr.19: 45590866-45590866 on Build GRCh38
rs200530342 Chr.19: 45590837-45590837 on Build GRCh38
rs36028976 Chr.19: 45590823-45590823 on Build GRCh38
rs746124143 Chr.19: 45590829-45590829 on Build GRCh38
rs201440587 Chr.19: 45590792-45590792 on Build GRCh38
rs769783412 Chr.19: 45590836-45590836 on Build GRCh38
rs1006726644 Chr.19: 45590676-45590676 on Build GRCh38
rs758035724 Chr.19: 45590773-45590773 on Build GRCh38
rs761788269 Chr.19: 45590743-45590743 on Build GRCh38
rs776456161 Chr.19: 45590698-45590698 on Build GRCh38
rs759467890 Chr.19: 45590730-45590730 on Build GRCh38
rs766931648 Chr.19: 45590872-45590872 on Build GRCh38
rs375682884 Chr.19: 45590736-45590736 on Build GRCh38
rs775682837 Chr.19: 45590832-45590832 on Build GRCh38
rs752313186 Chr.19: 45590877-45590877 on Build GRCh38
rs996873671 Chr.19: 45590674-45590674 on Build GRCh38
rs754393335 Chr.19: 45590785-45590785 on Build GRCh38
rs770537988 Chr.19: 45590686-45590686 on Build GRCh38
rs765892081 Chr.19: 45590776-45590776 on Build GRCh38
rs778965698 Chr.19: 45590827-45590827 on Build GRCh38
rs945031275 Chr.19: 45590816-45590816 on Build GRCh38
rs749605411 Chr.19: 45590813-45590813 on Build GRCh38
rs1030325471 Chr.19: 45590706-45590706 on Build GRCh38
rs750288781 Chr.19: 45590748-45590748 on Build GRCh38
rs766216041 Chr.19: 45590840-45590840 on Build GRCh38

View Full Product Details