Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 25563610-25563839 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LDLRAP1
Entrez Gene ID
26119
Gene Name
low density lipoprotein receptor adaptor protein 1
Gene Aliases
ARH, ARH1, ARH2, FHCB1, FHCB2
UniGene
Hs.590911
Gene Chromosome Location
Chr.1: 25543580-25590400 on Build GRCh38
Transcripts
NM_015627.2 XM_006710559.3 XM_006710560.3 XM_006710561.3 XM_011541209.2 XM_011541210.2 XM_011541211.2 XM_011541212.2 XM_017000994.1 XM_017000995.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6700344 c.617-1G>A p.? Chr.1: 25563660-25563660 on Build GRCh38
3689539 c.675G>A p.T225T Chr.1: 25563719-25563719 on Build GRCh38
7295128 c.714G>A p.R238R Chr.1: 25563758-25563758 on Build GRCh38
3711305 c.618G>T p.L206F Chr.1: 25563662-25563662 on Build GRCh38
4504429 c.664C>T p.P222S Chr.1: 25563708-25563708 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375092296 Chr.1: 25563652-25563652 on Build GRCh38
rs768813208 Chr.1: 25563764-25563764 on Build GRCh38
rs752255281 Chr.1: 25563718-25563718 on Build GRCh38
rs201935533 Chr.1: 25563639-25563639 on Build GRCh38
rs757653305 Chr.1: 25563747-25563747 on Build GRCh38
rs746229586 Chr.1: 25563754-25563754 on Build GRCh38
rs146122441 Chr.1: 25563666-25563666 on Build GRCh38
rs936665483 Chr.1: 25563773-25563773 on Build GRCh38
rs759304895 Chr.1: 25563812-25563812 on Build GRCh38
rs926407411 Chr.1: 25563719-25563719 on Build GRCh38
rs767484345 Chr.1: 25563817-25563817 on Build GRCh38
rs386629679 Chr.1: 25563697-25563698 on Build GRCh38
rs554028072 Chr.1: 25563665-25563665 on Build GRCh38
rs748256604 Chr.1: 25563775-25563775 on Build GRCh38
rs542234892 Chr.1: 25563709-25563709 on Build GRCh38
rs780616142 Chr.1: 25563640-25563640 on Build GRCh38
rs138552317 Chr.1: 25563749-25563749 on Build GRCh38
rs141522360 Chr.1: 25563670-25563670 on Build GRCh38
rs762630502 Chr.1: 25563632-25563632 on Build GRCh38
rs41291054 Chr.1: 25563716-25563716 on Build GRCh38
rs148579379 Chr.1: 25563757-25563757 on Build GRCh38
rs41291058 Chr.1: 25563756-25563756 on Build GRCh38
rs868577074 Chr.1: 25563802-25563802 on Build GRCh38
rs146324847 Chr.1: 25563647-25563647 on Build GRCh38
rs28969504 Chr.1: 25563698-25563698 on Build GRCh38
rs766377176 Chr.1: 25563634-25563634 on Build GRCh38
rs767432973 Chr.1: 25563717-25563717 on Build GRCh38
rs112914959 Chr.1: 25563755-25563755 on Build GRCh38
rs774075033 Chr.1: 25563807-25563807 on Build GRCh38
rs1056123624 Chr.1: 25563732-25563732 on Build GRCh38
rs143563728 Chr.1: 25563720-25563720 on Build GRCh38
rs753384633 Chr.1: 25563741-25563741 on Build GRCh38
rs746302717 Chr.1: 25563653-25563653 on Build GRCh38
rs114583297 Chr.1: 25563697-25563697 on Build GRCh38
rs776681796 Chr.1: 25563689-25563689 on Build GRCh38
rs549476432 Chr.1: 25563648-25563648 on Build GRCh38
rs533527945 Chr.1: 25563766-25563766 on Build GRCh38
rs202247605 Chr.1: 25563804-25563804 on Build GRCh38
rs375096767 Chr.1: 25563820-25563820 on Build GRCh38
rs140901487 Chr.1: 25563725-25563725 on Build GRCh38
rs373225679 Chr.1: 25563818-25563818 on Build GRCh38

View Full Product Details