Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 87590056-87590287 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ORC3
Entrez Gene ID
23595
Gene Name
origin recognition complex subunit 3
Gene Aliases
LAT, LATHEO, ORC3L
UniGene
Hs.410228
Gene Chromosome Location
Chr.6: 87590067-87677824 on Build GRCh38
Transcripts
NM_001197259.1 NM_012381.3 NM_181837.2 XM_005248704.1 XM_005248705.1 XM_005248706.1 XM_011535651.1 XM_011535652.2 XM_017010632.1 XM_017010633.1 XM_017010634.1 XM_017010635.1 XM_017010636.1 XM_017010637.1 XM_017010638.1 XM_017010639.1 XM_017010640.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1754950 c.14C>T p.S5L Chr.6: 87590182-87590182 on Build GRCh38
330514 c.10T>C p.S4P Chr.6: 87590178-87590178 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs750447344 Chr.6: 87590124-87590124 on Build GRCh38
rs757119067 Chr.6: 87590185-87590185 on Build GRCh38
rs767029921 Chr.6: 87590128-87590128 on Build GRCh38
rs774400857 Chr.6: 87590224-87590224 on Build GRCh38
rs896188778 Chr.6: 87590079-87590079 on Build GRCh38
rs2307368 Chr.6: 87590167-87590167 on Build GRCh38
rs766390045 Chr.6: 87590168-87590168 on Build GRCh38
rs1014427264 Chr.6: 87590080-87590080 on Build GRCh38
rs537955073 Chr.6: 87590149-87590149 on Build GRCh38
rs1043191787 Chr.6: 87590106-87590106 on Build GRCh38
rs867299625 Chr.6: 87590154-87590155 on Build GRCh38
rs766474607 Chr.6: 87590127-87590127 on Build GRCh38
rs986114751 Chr.6: 87590077-87590077 on Build GRCh38
rs554148837 Chr.6: 87590213-87590213 on Build GRCh38
rs2307367 Chr.6: 87590157-87590158 on Build GRCh38
rs924702560 Chr.6: 87590132-87590132 on Build GRCh38
rs750069069 Chr.6: 87590148-87590149 on Build GRCh38
rs781742133 Chr.6: 87590205-87590205 on Build GRCh38
rs2307387 Chr.6: 87590087-87590087 on Build GRCh38
rs761984082 Chr.6: 87590094-87590094 on Build GRCh38
rs918500366 Chr.6: 87590169-87590169 on Build GRCh38
rs777478399 Chr.6: 87590135-87590135 on Build GRCh38
rs756041517 Chr.6: 87590176-87590176 on Build GRCh38
rs200739084 Chr.6: 87590214-87590214 on Build GRCh38
rs748632871 Chr.6: 87590212-87590212 on Build GRCh38
rs28381461 Chr.6: 87590156-87590157 on Build GRCh38
rs375214415 Chr.6: 87590182-87590182 on Build GRCh38
rs758126071 Chr.6: 87590197-87590197 on Build GRCh38
rs929635001 Chr.6: 87590178-87590178 on Build GRCh38
rs777136487 Chr.6: 87590119-87590119 on Build GRCh38
rs752584358 Chr.6: 87590133-87590133 on Build GRCh38
rs771210155 Chr.6: 87590220-87590220 on Build GRCh38
rs556668324 Chr.6: 87590160-87590160 on Build GRCh38
rs776845912 Chr.6: 87590159-87590159 on Build GRCh38
rs539058759 Chr.6: 87590204-87590204 on Build GRCh38
rs1000430516 Chr.6: 87590091-87590091 on Build GRCh38
rs1056259657 Chr.6: 87590075-87590075 on Build GRCh38
rs765344789 Chr.6: 87590165-87590165 on Build GRCh38
rs758544701 Chr.6: 87590126-87590126 on Build GRCh38
rs146601654 Chr.6: 87590101-87590101 on Build GRCh38
rs761932297 Chr.6: 87590120-87590120 on Build GRCh38
rs751643866 Chr.6: 87590175-87590175 on Build GRCh38
rs906126096 Chr.6: 87590088-87590088 on Build GRCh38
rs149941248 Chr.6: 87590186-87590186 on Build GRCh38
rs778606342 Chr.6: 87590192-87590192 on Build GRCh38
rs1044063642 Chr.6: 87590174-87590174 on Build GRCh38
rs201438682 Chr.6: 87590134-87590134 on Build GRCh38
rs985326763 Chr.6: 87590144-87590144 on Build GRCh38
rs779702385 Chr.6: 87590199-87590199 on Build GRCh38
rs1018554682 Chr.6: 87590166-87590166 on Build GRCh38
rs771671266 Chr.6: 87590152-87590152 on Build GRCh38
rs747450079 Chr.6: 87590154-87590154 on Build GRCh38
rs746571124 Chr.6: 87590203-87590203 on Build GRCh38
rs376418108 Chr.6: 87590180-87590180 on Build GRCh38
rs750032137 Chr.6: 87590195-87590195 on Build GRCh38
rs548001675 Chr.6: 87590108-87590108 on Build GRCh38
rs750700001 Chr.6: 87590102-87590102 on Build GRCh38
rs765984236 Chr.6: 87590162-87590163 on Build GRCh38
rs758610491 Chr.6: 87590156-87590156 on Build GRCh38
rs759666253 Chr.6: 87590240-87590240 on Build GRCh38
rs756850039 Chr.6: 87590136-87590136 on Build GRCh38

View Full Product Details