Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 80144753-80145014 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLAC9
Entrez Gene ID
219348
Gene Name
placenta specific 9
Gene Aliases
-
UniGene
Hs.204947
Gene Chromosome Location
Chr.10: 80132502-80145028 on Build GRCh38
Transcripts
NM_001012973.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3981158 c.284-6T>G p.? Chr.10: 80144894-80144894 on Build GRCh38
7231085 c.223C>T p.R75C Chr.10: 80144960-80144960 on Build GRCh38
3981159 c.163-6T>G p.? Chr.10: 80144894-80144894 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs993554989 Chr.10: 80144940-80144940 on Build GRCh38
rs376212639 Chr.10: 80144913-80144913 on Build GRCh38
rs375397586 Chr.10: 80144936-80144936 on Build GRCh38
rs754226546 Chr.10: 80144917-80144917 on Build GRCh38
rs2819874 Chr.10: 80144882-80144882 on Build GRCh38
rs749007604 Chr.10: 80144857-80144857 on Build GRCh38
rs115345620 Chr.10: 80144987-80144987 on Build GRCh38
rs778582239 Chr.10: 80144919-80144919 on Build GRCh38
rs143576526 Chr.10: 80144786-80144786 on Build GRCh38
rs370749701 Chr.10: 80144918-80144918 on Build GRCh38
rs771929785 Chr.10: 80144878-80144878 on Build GRCh38
rs377049446 Chr.10: 80144847-80144847 on Build GRCh38
rs533076289 Chr.10: 80144776-80144776 on Build GRCh38
rs756481160 Chr.10: 80144850-80144850 on Build GRCh38
rs555422167 Chr.10: 80144903-80144903 on Build GRCh38
rs1129746 Chr.10: 80144907-80144907 on Build GRCh38
rs756951360 Chr.10: 80144858-80144858 on Build GRCh38
rs746807160 Chr.10: 80144880-80144880 on Build GRCh38
rs972251934 Chr.10: 80144783-80144783 on Build GRCh38
rs779980688 Chr.10: 80144879-80144879 on Build GRCh38
rs768556505 Chr.10: 80144980-80144980 on Build GRCh38
rs774326331 Chr.10: 80144897-80144897 on Build GRCh38
rs761252247 Chr.10: 80144912-80144912 on Build GRCh38
rs534904509 Chr.10: 80144824-80144824 on Build GRCh38
rs570733113 Chr.10: 80144840-80144840 on Build GRCh38
rs778111722 Chr.10: 80144837-80144837 on Build GRCh38
rs138810772 Chr.10: 80144901-80144901 on Build GRCh38
rs1027814117 Chr.10: 80144784-80144784 on Build GRCh38
rs549154404 Chr.10: 80144810-80144810 on Build GRCh38
rs778330049 Chr.10: 80144864-80144864 on Build GRCh38
rs913691340 Chr.10: 80144812-80144816 on Build GRCh38
rs937947219 Chr.10: 80144870-80144870 on Build GRCh38
rs758534666 Chr.10: 80144785-80144785 on Build GRCh38
rs745347686 Chr.10: 80144872-80144872 on Build GRCh38
rs868096843 Chr.10: 80144943-80144943 on Build GRCh38
rs41291392 Chr.10: 80144889-80144889 on Build GRCh38
rs148022488 Chr.10: 80144808-80144808 on Build GRCh38
rs748501446 Chr.10: 80144789-80144789 on Build GRCh38
rs113048746 Chr.10: 80144809-80144809 on Build GRCh38
rs539791817 Chr.10: 80144881-80144881 on Build GRCh38
rs779422794 Chr.10: 80144974-80144974 on Build GRCh38
rs749910603 Chr.10: 80144935-80144935 on Build GRCh38
rs770623138 Chr.10: 80144911-80144911 on Build GRCh38
rs778280121 Chr.10: 80144852-80144852 on Build GRCh38
rs547801698 Chr.10: 80144838-80144838 on Build GRCh38
rs566023932 Chr.10: 80144813-80144813 on Build GRCh38
rs747004796 Chr.10: 80144979-80144979 on Build GRCh38
rs867137924 Chr.10: 80144925-80144925 on Build GRCh38
rs747561092 Chr.10: 80144807-80144807 on Build GRCh38

View Full Product Details