Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 116941828-116942101 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTGFRN
Entrez Gene ID
5738
Gene Name
prostaglandin F2 receptor inhibitor
Gene Aliases
CD315, CD9P-1, EWI-F, FPRP, SMAP-6
UniGene
Hs.418093
Gene Chromosome Location
Chr.1: 116909923-116990358 on Build GRCh38
Transcripts
NM_020440.3 XM_017001874.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6361608 c.232T>G p.F78V Chr.1: 116941897-116941897 on Build GRCh38
5511561 c.256C>T p.R86W Chr.1: 116941921-116941921 on Build GRCh38
894452 c.377C>T p.A126V Chr.1: 116942042-116942042 on Build GRCh38
5799689 c.296A>G p.N99S Chr.1: 116941961-116941961 on Build GRCh38
6758490 c.257G>A p.R86Q Chr.1: 116941922-116941922 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs999372120 Chr.1: 116941953-116941953 on Build GRCh38
rs774022293 Chr.1: 116941961-116941961 on Build GRCh38
rs768459857 Chr.1: 116942060-116942060 on Build GRCh38
rs890473558 Chr.1: 116942024-116942024 on Build GRCh38
rs778728062 Chr.1: 116942061-116942061 on Build GRCh38
rs767176376 Chr.1: 116941963-116941963 on Build GRCh38
rs77895792 Chr.1: 116941959-116941959 on Build GRCh38
rs149801733 Chr.1: 116941962-116941962 on Build GRCh38
rs745989853 Chr.1: 116942005-116942005 on Build GRCh38
rs779856725 Chr.1: 116942043-116942043 on Build GRCh38
rs772638079 Chr.1: 116941965-116941965 on Build GRCh38
rs749181213 Chr.1: 116942047-116942047 on Build GRCh38
rs749091515 Chr.1: 116941956-116941956 on Build GRCh38
rs372565948 Chr.1: 116941981-116941981 on Build GRCh38
rs757606870 Chr.1: 116941989-116941989 on Build GRCh38
rs753070067 Chr.1: 116941883-116941883 on Build GRCh38
rs539247722 Chr.1: 116941974-116941974 on Build GRCh38
rs111346071 Chr.1: 116941943-116941943 on Build GRCh38
rs772905684 Chr.1: 116942075-116942075 on Build GRCh38
rs1024808076 Chr.1: 116941892-116941892 on Build GRCh38
rs374330870 Chr.1: 116942019-116942019 on Build GRCh38
rs757446946 Chr.1: 116941918-116941918 on Build GRCh38
rs781086898 Chr.1: 116941926-116941926 on Build GRCh38
rs758840988 Chr.1: 116941975-116941975 on Build GRCh38
rs747163742 Chr.1: 116941911-116941911 on Build GRCh38
rs970992669 Chr.1: 116941930-116941930 on Build GRCh38
rs775480414 Chr.1: 116941941-116941941 on Build GRCh38
rs778155592 Chr.1: 116941907-116941907 on Build GRCh38
rs764516019 Chr.1: 116941980-116941980 on Build GRCh38
rs368615013 Chr.1: 116941968-116941968 on Build GRCh38
rs368215880 Chr.1: 116942062-116942062 on Build GRCh38
rs771688649 Chr.1: 116942068-116942068 on Build GRCh38
rs201525760 Chr.1: 116941937-116941937 on Build GRCh38
rs765744080 Chr.1: 116941860-116941860 on Build GRCh38
rs745942048 Chr.1: 116941935-116941935 on Build GRCh38
rs769663692 Chr.1: 116941936-116941936 on Build GRCh38
rs1012614038 Chr.1: 116941867-116941867 on Build GRCh38
rs781449023 Chr.1: 116941990-116941990 on Build GRCh38
rs765722452 Chr.1: 116941969-116941969 on Build GRCh38
rs758822929 Chr.1: 116941893-116941893 on Build GRCh38

View Full Product Details