Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 19110778-19111005 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC25A42
Entrez Gene ID
284439
Gene Name
solute carrier family 25 member 42
Gene Aliases
-
UniGene
Hs.303669
Gene Chromosome Location
Chr.19: 19063961-19113030 on Build GRCh38
Transcripts
NM_001321544.1 NM_178526.4 XM_005259861.3 XM_011527937.2 XM_017026670.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
460014 c.864G>C p.L288F Chr.19: 19110783-19110783 on Build GRCh38
6780746 c.922C>A p.L308I Chr.19: 19110841-19110841 on Build GRCh38
3668211 c.868A>T p.M290L Chr.19: 19110787-19110787 on Build GRCh38
6390495 c.919G>A p.D307N Chr.19: 19110838-19110838 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs781548124 Chr.19: 19110864-19110864 on Build GRCh38
rs773324170 Chr.19: 19110908-19110908 on Build GRCh38
rs910099620 Chr.19: 19110942-19110942 on Build GRCh38
rs762744614 Chr.19: 19110926-19110926 on Build GRCh38
rs571547362 Chr.19: 19110969-19110969 on Build GRCh38
rs1002724033 Chr.19: 19110972-19110972 on Build GRCh38
rs767612799 Chr.19: 19110798-19110798 on Build GRCh38
rs1041667656 Chr.19: 19110962-19110962 on Build GRCh38
rs746346224 Chr.19: 19110890-19110890 on Build GRCh38
rs908992952 Chr.19: 19110887-19110887 on Build GRCh38
rs188474907 Chr.19: 19110898-19110898 on Build GRCh38
rs757977417 Chr.19: 19110852-19110852 on Build GRCh38
rs759515020 Chr.19: 19110828-19110828 on Build GRCh38
rs893260667 Chr.19: 19110945-19110945 on Build GRCh38
rs757354752 Chr.19: 19110901-19110901 on Build GRCh38
rs958957070 Chr.19: 19110944-19110944 on Build GRCh38
rs551555864 Chr.19: 19110934-19110934 on Build GRCh38
rs749606569 Chr.19: 19110897-19110897 on Build GRCh38
rs762041639 Chr.19: 19110901-19110901 on Build GRCh38
rs958585630 Chr.19: 19110855-19110855 on Build GRCh38
rs756410811 Chr.19: 19110946-19110946 on Build GRCh38
rs753722901 Chr.19: 19110825-19110825 on Build GRCh38
rs554978947 Chr.19: 19110943-19110944 on Build GRCh38
rs529307843 Chr.19: 19110860-19110860 on Build GRCh38
rs759469967 Chr.19: 19110917-19110917 on Build GRCh38
rs780516298 Chr.19: 19110967-19110967 on Build GRCh38
rs780871965 Chr.19: 19110879-19110879 on Build GRCh38
rs752608316 Chr.19: 19110851-19110851 on Build GRCh38
rs984716237 Chr.19: 19110932-19110932 on Build GRCh38
rs17854358 Chr.19: 19110853-19110853 on Build GRCh38
rs780267496 Chr.19: 19110896-19110896 on Build GRCh38
rs772113390 Chr.19: 19110905-19110905 on Build GRCh38
rs752593495 Chr.19: 19110918-19110918 on Build GRCh38
rs1012078691 Chr.19: 19110953-19110953 on Build GRCh38
rs549374233 Chr.19: 19110893-19110893 on Build GRCh38
rs140099041 Chr.19: 19110830-19110830 on Build GRCh38

View Full Product Details