Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 13768240-13768513 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MRI1
Entrez Gene ID
84245
Gene Name
methylthioribose-1-phosphate isomerase 1
Gene Aliases
MRDI, MTNA, Ypr118w
UniGene
Hs.439370
Gene Chromosome Location
Chr.19: 13764492-13774282 on Build GRCh38
Transcripts
NM_001031727.2 NM_032285.2 XM_011528355.2 XM_011528356.2 XM_011528357.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5513478 c.433C>T p.R145* Chr.19: 13768420-13768420 on Build GRCh38
6281732 c.391G>A p.E131K Chr.19: 13768378-13768378 on Build GRCh38
6367196 c.392A>G p.E131G Chr.19: 13768379-13768379 on Build GRCh38
211858 c.403G>A p.E135K Chr.19: 13768390-13768390 on Build GRCh38
991734 c.441C>G p.P147P Chr.19: 13768428-13768428 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777633007 Chr.19: 13768272-13768272 on Build GRCh38
rs746642716 Chr.19: 13768291-13768291 on Build GRCh38
rs866810807 Chr.19: 13768362-13768362 on Build GRCh38
rs148498603 Chr.19: 13768421-13768421 on Build GRCh38
rs760933462 Chr.19: 13768419-13768419 on Build GRCh38
rs768076933 Chr.19: 13768479-13768479 on Build GRCh38
rs868765438 Chr.19: 13768455-13768455 on Build GRCh38
rs900049892 Chr.19: 13768344-13768344 on Build GRCh38
rs867590578 Chr.19: 13768412-13768412 on Build GRCh38
rs146793123 Chr.19: 13768377-13768377 on Build GRCh38
rs375787163 Chr.19: 13768435-13768435 on Build GRCh38
rs941059490 Chr.19: 13768315-13768315 on Build GRCh38
rs374680191 Chr.19: 13768468-13768468 on Build GRCh38
rs749752463 Chr.19: 13768449-13768449 on Build GRCh38
rs771529455 Chr.19: 13768455-13768455 on Build GRCh38
rs771477499 Chr.19: 13768372-13768372 on Build GRCh38
rs949881347 Chr.19: 13768346-13768346 on Build GRCh38
rs776258287 Chr.19: 13768378-13768378 on Build GRCh38
rs750287203 Chr.19: 13768434-13768434 on Build GRCh38
rs372934503 Chr.19: 13768338-13768338 on Build GRCh38
rs766282290 Chr.19: 13768437-13768437 on Build GRCh38
rs770894668 Chr.19: 13768302-13768302 on Build GRCh38
rs368348656 Chr.19: 13768337-13768337 on Build GRCh38
rs1007586012 Chr.19: 13768490-13768490 on Build GRCh38
rs758722180 Chr.19: 13768464-13768464 on Build GRCh38
rs991968548 Chr.19: 13768438-13768438 on Build GRCh38
rs369252426 Chr.19: 13768446-13768446 on Build GRCh38
rs768906445 Chr.19: 13768320-13768320 on Build GRCh38
rs376290350 Chr.19: 13768300-13768300 on Build GRCh38
rs184093445 Chr.19: 13768474-13768474 on Build GRCh38
rs748777027 Chr.19: 13768475-13768475 on Build GRCh38
rs773891174 Chr.19: 13768383-13768383 on Build GRCh38
rs753944966 Chr.19: 13768442-13768442 on Build GRCh38
rs1047207293 Chr.19: 13768318-13768318 on Build GRCh38
rs142646684 Chr.19: 13768447-13768447 on Build GRCh38
rs748391191 Chr.19: 13768453-13768453 on Build GRCh38
rs773659295 Chr.19: 13768361-13768361 on Build GRCh38
rs747545091 Chr.19: 13768368-13768368 on Build GRCh38
rs903552392 Chr.19: 13768420-13768420 on Build GRCh38
rs771424740 Chr.19: 13768469-13768469 on Build GRCh38
rs776009461 Chr.19: 13768429-13768438 on Build GRCh38
rs769567780 Chr.19: 13768417-13768417 on Build GRCh38
rs191399666 Chr.19: 13768292-13768292 on Build GRCh38
rs1045574385 Chr.19: 13768371-13768371 on Build GRCh38
rs796672268 Chr.19: 13768336-13768336 on Build GRCh38
rs778088741 Chr.19: 13768467-13768467 on Build GRCh38
rs772437959 Chr.19: 13768360-13768360 on Build GRCh38
rs541895914 Chr.19: 13768379-13768379 on Build GRCh38
rs745848142 Chr.19: 13768385-13768385 on Build GRCh38

View Full Product Details