Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 75005771-75006026 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NEU3
Entrez Gene ID
10825
Gene Name
neuraminidase 3
Gene Aliases
SIAL3
UniGene
Hs.191074
Gene Chromosome Location
Chr.11: 74988905-75007698 on Build GRCh38
Transcripts
NM_006656.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5775348 c.901C>A p.L301M Chr.11: 75006007-75006007 on Build GRCh38
1217119 c.832C>T p.R278W Chr.11: 75005938-75005938 on Build GRCh38
6724753 c.855A>G p.A285A Chr.11: 75005961-75005961 on Build GRCh38
1259019 c.780A>T p.E260D Chr.11: 75005886-75005886 on Build GRCh38
4902511 c.700C>T p.L234L Chr.11: 75005806-75005806 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781223780 Chr.11: 75005996-75005996 on Build GRCh38
rs767782339 Chr.11: 75005866-75005866 on Build GRCh38
rs376361957 Chr.11: 75005938-75005938 on Build GRCh38
rs779067414 Chr.11: 75005914-75005914 on Build GRCh38
rs759130871 Chr.11: 75005829-75005829 on Build GRCh38
rs755089060 Chr.11: 75005911-75005911 on Build GRCh38
rs778122110 Chr.11: 75005987-75005987 on Build GRCh38
rs770766955 Chr.11: 75005928-75005928 on Build GRCh38
rs763867572 Chr.11: 75005949-75005949 on Build GRCh38
rs200007739 Chr.11: 75005957-75005957 on Build GRCh38
rs371627063 Chr.11: 75005812-75005812 on Build GRCh38
rs767180765 Chr.11: 75005829-75005830 on Build GRCh38
rs770409657 Chr.11: 75005823-75005823 on Build GRCh38
rs775221082 Chr.11: 75005940-75005940 on Build GRCh38
rs17133599 Chr.11: 75005925-75005925 on Build GRCh38
rs753890800 Chr.11: 75005909-75005909 on Build GRCh38
rs760544688 Chr.11: 75005819-75005819 on Build GRCh38
rs906621867 Chr.11: 75005878-75005878 on Build GRCh38
rs897785622 Chr.11: 75005933-75005933 on Build GRCh38
rs147894902 Chr.11: 75005829-75005829 on Build GRCh38
rs944512057 Chr.11: 75005935-75005935 on Build GRCh38
rs1045146884 Chr.11: 75005867-75005867 on Build GRCh38
rs759777638 Chr.11: 75005868-75005868 on Build GRCh38
rs752858195 Chr.11: 75005977-75005977 on Build GRCh38
rs777441967 Chr.11: 75005923-75005923 on Build GRCh38
rs867312992 Chr.11: 75005901-75005902 on Build GRCh38
rs539514716 Chr.11: 75005956-75005956 on Build GRCh38
rs776747427 Chr.11: 75005936-75005936 on Build GRCh38
rs769447064 Chr.11: 75005939-75005939 on Build GRCh38
rs369776172 Chr.11: 75005849-75005849 on Build GRCh38
rs773153256 Chr.11: 75005818-75005818 on Build GRCh38
rs748401241 Chr.11: 75005917-75005917 on Build GRCh38
rs200629627 Chr.11: 75005890-75005890 on Build GRCh38
rs915421795 Chr.11: 75005833-75005833 on Build GRCh38
rs766775465 Chr.11: 75005960-75005960 on Build GRCh38
rs765305268 Chr.11: 75005976-75005976 on Build GRCh38
rs762233174 Chr.11: 75005863-75005863 on Build GRCh38
rs1001769382 Chr.11: 75005975-75005975 on Build GRCh38
rs780666537 Chr.11: 75005904-75005904 on Build GRCh38
rs771832431 Chr.11: 75005919-75005919 on Build GRCh38
rs1043777997 Chr.11: 75005963-75005963 on Build GRCh38
rs527628861 Chr.11: 75005855-75005855 on Build GRCh38
rs548397936 Chr.11: 75005814-75005814 on Build GRCh38
rs758534119 Chr.11: 75005982-75005982 on Build GRCh38
rs747906258 Chr.11: 75005800-75005800 on Build GRCh38
rs750872545 Chr.11: 75005886-75005886 on Build GRCh38
rs201144132 Chr.11: 75005992-75005992 on Build GRCh38
rs373455752 Chr.11: 75005989-75005989 on Build GRCh38
rs369520041 Chr.11: 75005943-75005943 on Build GRCh38
rs201661038 Chr.11: 75005966-75005966 on Build GRCh38
rs904815986 Chr.11: 75005971-75005971 on Build GRCh38
rs752354749 Chr.11: 75006005-75006005 on Build GRCh38

View Full Product Details