Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1089467-1089712 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POLR2E
Entrez Gene ID
5434
Gene Name
RNA polymerase II subunit E
Gene Aliases
RPABC1, RPB5, XAP4, hRPB25, hsRPB5
UniGene
Hs.24301
Gene Chromosome Location
Chr.19: 1086579-1095392 on Build GRCh38
Transcripts
NM_001316323.1 NM_001316324.1 NM_002695.3 XM_011528070.2 XM_017026883.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4131176 c.626T>G p.V209G Chr.19: 1089493-1089493 on Build GRCh38
3932869 c.592G>A p.E198K Chr.19: 1089527-1089527 on Build GRCh38
3959499 c.568-6C>T p.? Chr.19: 1089557-1089557 on Build GRCh38
396993 c.601G>A p.G201S Chr.19: 1089518-1089518 on Build GRCh38
2813165 c.603C>T p.G201G Chr.19: 1089516-1089516 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749816315 Chr.19: 1089528-1089528 on Build GRCh38
rs200508955 Chr.19: 1089589-1089589 on Build GRCh38
rs1036915394 Chr.19: 1089621-1089621 on Build GRCh38
rs368638025 Chr.19: 1089593-1089593 on Build GRCh38
rs533403115 Chr.19: 1089685-1089685 on Build GRCh38
rs958008404 Chr.19: 1089667-1089667 on Build GRCh38
rs780622181 Chr.19: 1089523-1089523 on Build GRCh38
rs889562284 Chr.19: 1089639-1089639 on Build GRCh38
rs950570466 Chr.19: 1089635-1089635 on Build GRCh38
rs560882875 Chr.19: 1089644-1089644 on Build GRCh38
rs756854412 Chr.19: 1089580-1089580 on Build GRCh38
rs200847033 Chr.19: 1089578-1089578 on Build GRCh38
rs772396366 Chr.19: 1089553-1089553 on Build GRCh38
rs967342267 Chr.19: 1089488-1089488 on Build GRCh38
rs939901146 Chr.19: 1089602-1089602 on Build GRCh38
rs766825144 Chr.19: 1089686-1089686 on Build GRCh38
rs546866585 Chr.19: 1089656-1089656 on Build GRCh38
rs773489108 Chr.19: 1089588-1089588 on Build GRCh38
rs763171549 Chr.19: 1089636-1089636 on Build GRCh38
rs772154822 Chr.19: 1089486-1089486 on Build GRCh38
rs773392791 Chr.19: 1089489-1089489 on Build GRCh38
rs760916464 Chr.19: 1089556-1089556 on Build GRCh38
rs748440861 Chr.19: 1089547-1089547 on Build GRCh38
rs760824411 Chr.19: 1089490-1089490 on Build GRCh38
rs370150594 Chr.19: 1089535-1089535 on Build GRCh38
rs990902751 Chr.19: 1089570-1089570 on Build GRCh38
rs779277090 Chr.19: 1089591-1089591 on Build GRCh38
rs752108811 Chr.19: 1089691-1089691 on Build GRCh38
rs199507033 Chr.19: 1089565-1089565 on Build GRCh38
rs765060604 Chr.19: 1089576-1089576 on Build GRCh38
rs765106012 Chr.19: 1089501-1089501 on Build GRCh38
rs921226010 Chr.19: 1089655-1089655 on Build GRCh38
rs752399749 Chr.19: 1089504-1089504 on Build GRCh38
rs112111019 Chr.19: 1089626-1089626 on Build GRCh38
rs146230282 Chr.19: 1089522-1089522 on Build GRCh38
rs766492286 Chr.19: 1089491-1089491 on Build GRCh38
rs372881876 Chr.19: 1089500-1089500 on Build GRCh38
rs911792456 Chr.19: 1089642-1089642 on Build GRCh38
rs996855359 Chr.19: 1089561-1089561 on Build GRCh38
rs563077325 Chr.19: 1089590-1089590 on Build GRCh38
rs1053802926 Chr.19: 1089563-1089563 on Build GRCh38
rs759581079 Chr.19: 1089574-1089574 on Build GRCh38
rs1049450593 Chr.19: 1089637-1089637 on Build GRCh38
rs759491281 Chr.19: 1089499-1089499 on Build GRCh38
rs191686722 Chr.19: 1089630-1089630 on Build GRCh38
rs973108815 Chr.19: 1089620-1089620 on Build GRCh38
rs777273136 Chr.19: 1089510-1089510 on Build GRCh38
rs758116915 Chr.19: 1089505-1089505 on Build GRCh38
rs776806763 Chr.19: 1089573-1089573 on Build GRCh38
rs777839600 Chr.19: 1089570-1089571 on Build GRCh38
rs746984062 Chr.19: 1089587-1089588 on Build GRCh38
rs375882249 Chr.19: 1089512-1089512 on Build GRCh38
rs759341193 Chr.19: 1089608-1089608 on Build GRCh38
rs1012284653 Chr.19: 1089684-1089684 on Build GRCh38
rs769002822 Chr.19: 1089533-1089533 on Build GRCh38
rs974641509 Chr.19: 1089651-1089651 on Build GRCh38
rs758141192 Chr.19: 1089562-1089563 on Build GRCh38
rs528394199 Chr.19: 1089645-1089645 on Build GRCh38
rs372411618 Chr.19: 1089555-1089555 on Build GRCh38
rs893912602 Chr.19: 1089665-1089665 on Build GRCh38
rs753834886 Chr.19: 1089493-1089493 on Build GRCh38
rs571559216 Chr.19: 1089661-1089661 on Build GRCh38
rs776771288 Chr.19: 1089599-1089599 on Build GRCh38
rs780709926 Chr.19: 1089584-1089584 on Build GRCh38
rs57575311 Chr.19: 1089577-1089577 on Build GRCh38

View Full Product Details