Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 5788116-5788389 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR52N1
Entrez Gene ID
79473
Gene Name
olfactory receptor family 52 subfamily N member 1
Gene Aliases
OR11-61
UniGene
Hs.690209
Gene Chromosome Location
Chr.11: 5787854-5788816 on Build GRCh38
Transcripts
NM_001001913.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3449926 c.670C>T p.L224F Chr.11: 5788147-5788147 on Build GRCh38
3449928 c.505C>T p.P169S Chr.11: 5788312-5788312 on Build GRCh38
3449927 c.558G>T p.V186V Chr.11: 5788259-5788259 on Build GRCh38
7381639 c.548A>T p.H183L Chr.11: 5788269-5788269 on Build GRCh38
3449929 c.502C>T p.L168F Chr.11: 5788315-5788315 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748249902 Chr.11: 5788249-5788249 on Build GRCh38
rs760078210 Chr.11: 5788198-5788198 on Build GRCh38
rs773631047 Chr.11: 5788179-5788179 on Build GRCh38
rs781188466 Chr.11: 5788306-5788306 on Build GRCh38
rs7948009 Chr.11: 5788318-5788318 on Build GRCh38
rs138814143 Chr.11: 5788361-5788361 on Build GRCh38
rs763409859 Chr.11: 5788186-5788186 on Build GRCh38
rs779334698 Chr.11: 5788343-5788343 on Build GRCh38
rs769042411 Chr.11: 5788358-5788358 on Build GRCh38
rs376517982 Chr.11: 5788360-5788360 on Build GRCh38
rs769835835 Chr.11: 5788255-5788255 on Build GRCh38
rs371764761 Chr.11: 5788310-5788310 on Build GRCh38
rs756095593 Chr.11: 5788317-5788317 on Build GRCh38
rs754790053 Chr.11: 5788305-5788305 on Build GRCh38
rs923707744 Chr.11: 5788282-5788282 on Build GRCh38
rs776042360 Chr.11: 5788357-5788357 on Build GRCh38
rs770002969 Chr.11: 5788178-5788178 on Build GRCh38
rs960914658 Chr.11: 5788180-5788180 on Build GRCh38
rs147301110 Chr.11: 5788225-5788225 on Build GRCh38
rs771307560 Chr.11: 5788266-5788266 on Build GRCh38
rs10838637 Chr.11: 5788247-5788247 on Build GRCh38
rs769182013 Chr.11: 5788359-5788359 on Build GRCh38
rs866716179 Chr.11: 5788183-5788183 on Build GRCh38
rs11823184 Chr.11: 5788175-5788175 on Build GRCh38
rs746839798 Chr.11: 5788235-5788235 on Build GRCh38
rs761479165 Chr.11: 5788193-5788193 on Build GRCh38
rs1034820225 Chr.11: 5788177-5788177 on Build GRCh38
rs887477307 Chr.11: 5788350-5788350 on Build GRCh38
rs774787785 Chr.11: 5788268-5788268 on Build GRCh38
rs758168503 Chr.11: 5788231-5788231 on Build GRCh38
rs749468499 Chr.11: 5788261-5788261 on Build GRCh38
rs768500128 Chr.11: 5788171-5788171 on Build GRCh38
rs764704251 Chr.11: 5788294-5788294 on Build GRCh38
rs752620306 Chr.11: 5788316-5788316 on Build GRCh38
rs139204244 Chr.11: 5788166-5788166 on Build GRCh38
rs368353863 Chr.11: 5788165-5788165 on Build GRCh38
rs746238148 Chr.11: 5788356-5788356 on Build GRCh38
rs779775471 Chr.11: 5788234-5788234 on Build GRCh38
rs1005231909 Chr.11: 5788142-5788142 on Build GRCh38
rs149811568 Chr.11: 5788339-5788339 on Build GRCh38
rs745646846 Chr.11: 5788156-5788156 on Build GRCh38
rs115404910 Chr.11: 5788301-5788301 on Build GRCh38
rs753026863 Chr.11: 5788333-5788333 on Build GRCh38
rs775928411 Chr.11: 5788285-5788285 on Build GRCh38
rs375501780 Chr.11: 5788209-5788209 on Build GRCh38
rs747000579 Chr.11: 5788167-5788167 on Build GRCh38
rs749380581 Chr.11: 5788328-5788328 on Build GRCh38
rs769148336 Chr.11: 5788192-5788192 on Build GRCh38
rs773543025 Chr.11: 5788258-5788258 on Build GRCh38
rs762555100 Chr.11: 5788300-5788300 on Build GRCh38
rs140764863 Chr.11: 5788226-5788226 on Build GRCh38
rs1014754975 Chr.11: 5788150-5788150 on Build GRCh38
rs754878934 Chr.11: 5788246-5788246 on Build GRCh38
rs749958918 Chr.11: 5788295-5788295 on Build GRCh38
rs768053820 Chr.11: 5788280-5788280 on Build GRCh38
rs866370287 Chr.11: 5788155-5788155 on Build GRCh38
rs761355183 Chr.11: 5788288-5788288 on Build GRCh38
rs866085693 Chr.11: 5788365-5788365 on Build GRCh38
rs753330009 Chr.11: 5788218-5788218 on Build GRCh38
rs767911405 Chr.11: 5788210-5788210 on Build GRCh38
rs774873910 Chr.11: 5788194-5788194 on Build GRCh38

View Full Product Details