Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 119838492-119838753 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MCMBP
Entrez Gene ID
79892
Gene Name
minichromosome maintenance complex binding protein
Gene Aliases
C10orf119, MCM-BP
UniGene
Hs.124246
Gene Chromosome Location
Chr.10: 119829404-119873628 on Build GRCh38
Transcripts
NM_001256378.1 NM_001256379.1 NM_024834.3 XM_005270157.4 XM_011540170.1 XM_017016663.1 XM_017016664.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1703153 c.1366C>T p.L456F Chr.10: 119838583-119838583 on Build GRCh38
4994839 c.1324C>T p.R442C Chr.10: 119838625-119838625 on Build GRCh38
1470298 c.1255C>T p.R419C Chr.10: 119838694-119838694 on Build GRCh38
3985455 c.1377T>C p.D459D Chr.10: 119838572-119838572 on Build GRCh38
5447991 c.1333A>G p.S445G Chr.10: 119838616-119838616 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766651867 Chr.10: 119838575-119838575 on Build GRCh38
rs200347354 Chr.10: 119838699-119838699 on Build GRCh38
rs933462887 Chr.10: 119838678-119838678 on Build GRCh38
rs933337288 Chr.10: 119838584-119838584 on Build GRCh38
rs759648667 Chr.10: 119838591-119838591 on Build GRCh38
rs202205398 Chr.10: 119838693-119838693 on Build GRCh38
rs1051951158 Chr.10: 119838694-119838694 on Build GRCh38
rs773796137 Chr.10: 119838711-119838734 on Build GRCh38
rs928421334 Chr.10: 119838516-119838516 on Build GRCh38
rs768173433 Chr.10: 119838552-119838552 on Build GRCh38
rs778463526 Chr.10: 119838670-119838670 on Build GRCh38
rs774798925 Chr.10: 119838565-119838565 on Build GRCh38
rs951100273 Chr.10: 119838671-119838671 on Build GRCh38
rs368405799 Chr.10: 119838559-119838559 on Build GRCh38
rs750163013 Chr.10: 119838627-119838627 on Build GRCh38
rs12772291 Chr.10: 119838661-119838661 on Build GRCh38
rs747668253 Chr.10: 119838679-119838679 on Build GRCh38
rs371212965 Chr.10: 119838548-119838548 on Build GRCh38
rs374240470 Chr.10: 119838555-119838555 on Build GRCh38
rs754136298 Chr.10: 119838585-119838585 on Build GRCh38
rs781060535 Chr.10: 119838625-119838625 on Build GRCh38
rs558673345 Chr.10: 119838719-119838719 on Build GRCh38
rs191720352 Chr.10: 119838530-119838530 on Build GRCh38
rs143374034 Chr.10: 119838690-119838690 on Build GRCh38
rs765380547 Chr.10: 119838607-119838607 on Build GRCh38
rs757208942 Chr.10: 119838623-119838623 on Build GRCh38
rs760954436 Chr.10: 119838574-119838574 on Build GRCh38
rs752771731 Chr.10: 119838611-119838611 on Build GRCh38
rs772503219 Chr.10: 119838567-119838567 on Build GRCh38
rs773611450 Chr.10: 119838572-119838572 on Build GRCh38
rs980712725 Chr.10: 119838563-119838563 on Build GRCh38
rs755914490 Chr.10: 119838630-119838630 on Build GRCh38
rs762350053 Chr.10: 119838566-119838566 on Build GRCh38
rs112748789 Chr.10: 119838717-119838717 on Build GRCh38
rs569672236 Chr.10: 119838581-119838581 on Build GRCh38
rs745710514 Chr.10: 119838531-119838531 on Build GRCh38
rs138353467 Chr.10: 119838634-119838634 on Build GRCh38
rs567849277 Chr.10: 119838521-119838521 on Build GRCh38
rs769429268 Chr.10: 119838538-119838538 on Build GRCh38
rs889342259 Chr.10: 119838708-119838708 on Build GRCh38
rs373697996 Chr.10: 119838525-119838525 on Build GRCh38
rs779683255 Chr.10: 119838547-119838547 on Build GRCh38
rs370286541 Chr.10: 119838522-119838522 on Build GRCh38
rs779629719 Chr.10: 119838633-119838633 on Build GRCh38

View Full Product Details