Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 74743938-74744199 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRPM6
Entrez Gene ID
140803
Gene Name
transient receptor potential cation channel subfamily M member 6
Gene Aliases
CHAK2, HMGX, HOMG, HOMG1, HSH
UniGene
Hs.272225
Gene Chromosome Location
Chr.9: 74722495-74888094 on Build GRCh38
Transcripts
NM_001177310.1 NM_001177311.1 NM_017662.4 XM_011518244.1 XM_011518245.1 XM_011518246.1 XM_011518247.1 XM_011518248.1 XM_011518249.1 XM_011518250.1 XM_011518251.2 XM_017014287.1 XM_017014288.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5411253 c.5088C>T p.I1696I Chr.9: 74744141-74744141 on Build GRCh38
6809723 c.5116A>G p.S1706G Chr.9: 74744125-74744125 on Build GRCh38
6809594 c.5105G>T p.S1702I Chr.9: 74744124-74744124 on Build GRCh38
7149461 c.5095C>G p.Q1699E Chr.9: 74744119-74744119 on Build GRCh38
6809725 c.5116A>G p.S1706G Chr.9: 74744125-74744125 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775951126 Chr.9: 74744056-74744056 on Build GRCh38
rs967851233 Chr.9: 74744025-74744025 on Build GRCh38
rs963952060 Chr.9: 74744087-74744087 on Build GRCh38
rs748124522 Chr.9: 74744072-74744072 on Build GRCh38
rs771038947 Chr.9: 74744163-74744163 on Build GRCh38
rs767669016 Chr.9: 74744124-74744124 on Build GRCh38
rs773201544 Chr.9: 74744077-74744077 on Build GRCh38
rs185242812 Chr.9: 74744074-74744074 on Build GRCh38
rs894606665 Chr.9: 74743991-74743991 on Build GRCh38
rs746540121 Chr.9: 74744051-74744051 on Build GRCh38
rs144525211 Chr.9: 74744053-74744053 on Build GRCh38
rs976983631 Chr.9: 74744117-74744117 on Build GRCh38
rs565181637 Chr.9: 74744007-74744007 on Build GRCh38
rs773933934 Chr.9: 74743990-74743990 on Build GRCh38
rs769785666 Chr.9: 74744076-74744076 on Build GRCh38
rs867161235 Chr.9: 74744104-74744104 on Build GRCh38
rs774611066 Chr.9: 74744085-74744085 on Build GRCh38
rs762840827 Chr.9: 74744082-74744082 on Build GRCh38
rs140493587 Chr.9: 74743993-74743993 on Build GRCh38
rs193058158 Chr.9: 74743992-74743992 on Build GRCh38
rs756625847 Chr.9: 74744164-74744164 on Build GRCh38
rs752709214 Chr.9: 74744136-74744136 on Build GRCh38
rs957955433 Chr.9: 74744058-74744058 on Build GRCh38
rs867452087 Chr.9: 74744122-74744122 on Build GRCh38
rs1055895466 Chr.9: 74743980-74743980 on Build GRCh38
rs761595871 Chr.9: 74744069-74744069 on Build GRCh38
rs575234607 Chr.9: 74744062-74744062 on Build GRCh38
rs766554589 Chr.9: 74744084-74744084 on Build GRCh38
rs759629217 Chr.9: 74744120-74744120 on Build GRCh38
rs557416219 Chr.9: 74743979-74743979 on Build GRCh38
rs764838440 Chr.9: 74744153-74744153 on Build GRCh38
rs989364206 Chr.9: 74744059-74744059 on Build GRCh38
rs998892907 Chr.9: 74744043-74744043 on Build GRCh38

View Full Product Details