Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 35552962-35553235 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNY
Entrez Gene ID
219771
Gene Name
cyclin Y
Gene Aliases
C10orf9, CBCP1, CCNX, CFP1
UniGene
Hs.14745
Gene Chromosome Location
Chr.10: 35246981-35572670 on Build GRCh38
Transcripts
NM_001282852.1 NM_001282853.1 NM_001282854.1 NM_145012.5 NM_181698.3 XM_006717392.3 XM_011519357.2 XM_011519358.1 XM_011519360.2 XM_011519361.1 XM_011519362.1 XM_011519363.1 XM_011519364.1 XM_011519365.2 XM_011519366.1 XM_011519367.1 XM_011519368.2 XM_011519369.2 XM_017015839.1 XM_017015840.1 XM_017015841.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6623121 c.641G>A p.R214Q Chr.10: 35553080-35553080 on Build GRCh38
6623120 c.530C>T p.A177V Chr.10: 35553131-35553131 on Build GRCh38
3807127 c.566A>C p.D189A Chr.10: 35553167-35553167 on Build GRCh38
3741643 c.494C>T p.A165V Chr.10: 35553095-35553095 on Build GRCh38
4013966 c.709T>C p.Y237H Chr.10: 35553148-35553148 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200117706 Chr.10: 35553085-35553085 on Build GRCh38
rs773480087 Chr.10: 35553069-35553069 on Build GRCh38
rs1014836264 Chr.10: 35553065-35553065 on Build GRCh38
rs753894235 Chr.10: 35553011-35553011 on Build GRCh38
rs752974472 Chr.10: 35553203-35553203 on Build GRCh38
rs764115191 Chr.10: 35553010-35553010 on Build GRCh38
rs536589641 Chr.10: 35553144-35553144 on Build GRCh38
rs866528929 Chr.10: 35553064-35553064 on Build GRCh38
rs747074559 Chr.10: 35553048-35553048 on Build GRCh38
rs147046743 Chr.10: 35553174-35553174 on Build GRCh38
rs752221237 Chr.10: 35553002-35553002 on Build GRCh38
rs768237252 Chr.10: 35553205-35553205 on Build GRCh38
rs770146084 Chr.10: 35553068-35553068 on Build GRCh38
rs150576108 Chr.10: 35553195-35553195 on Build GRCh38
rs373219386 Chr.10: 35553081-35553081 on Build GRCh38
rs776038975 Chr.10: 35553207-35553207 on Build GRCh38
rs764959644 Chr.10: 35553037-35553037 on Build GRCh38
rs761428879 Chr.10: 35553209-35553209 on Build GRCh38
rs765044750 Chr.10: 35553122-35553122 on Build GRCh38
rs369858323 Chr.10: 35553066-35553066 on Build GRCh38
rs377039217 Chr.10: 35552994-35552994 on Build GRCh38
rs370637331 Chr.10: 35553184-35553184 on Build GRCh38
rs376271533 Chr.10: 35553111-35553111 on Build GRCh38
rs923074279 Chr.10: 35553106-35553106 on Build GRCh38
rs781608774 Chr.10: 35553059-35553059 on Build GRCh38
rs576939183 Chr.10: 35553194-35553194 on Build GRCh38
rs765070593 Chr.10: 35553216-35553216 on Build GRCh38
rs377568126 Chr.10: 35553046-35553046 on Build GRCh38
rs761682655 Chr.10: 35553115-35553115 on Build GRCh38
rs867645681 Chr.10: 35553067-35553067 on Build GRCh38
rs751780823 Chr.10: 35553159-35553159 on Build GRCh38
rs369170090 Chr.10: 35553003-35553003 on Build GRCh38
rs556153316 Chr.10: 35553173-35553173 on Build GRCh38
rs200268976 Chr.10: 35553045-35553045 on Build GRCh38
rs749701305 Chr.10: 35553079-35553079 on Build GRCh38
rs757010312 Chr.10: 35553014-35553014 on Build GRCh38
rs746673282 Chr.10: 35553198-35553198 on Build GRCh38
rs758238578 Chr.10: 35552993-35552993 on Build GRCh38
rs142955158 Chr.10: 35553150-35553150 on Build GRCh38
rs750155432 Chr.10: 35553141-35553141 on Build GRCh38
rs372862951 Chr.10: 35553191-35553191 on Build GRCh38
rs35485034 Chr.10: 35553115-35553115 on Build GRCh38
rs767094283 Chr.10: 35552999-35552999 on Build GRCh38
rs911239190 Chr.10: 35553162-35553162 on Build GRCh38
rs755113583 Chr.10: 35553058-35553058 on Build GRCh38
rs763701965 Chr.10: 35553096-35553096 on Build GRCh38

View Full Product Details