Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 70672643-70672914 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADD2
Entrez Gene ID
119
Gene Name
adducin 2
Gene Aliases
ADDB
UniGene
Hs.188528
Gene Chromosome Location
Chr.2: 70656784-70768243 on Build GRCh38
Transcripts
NM_001185054.1 NM_001617.3 NM_017488.3 XM_011532502.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
297844 c.1862C>T p.S621F Chr.2: 70672886-70672886 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs782318291 Chr.2: 70672841-70672841 on Build GRCh38
rs6546617 Chr.2: 70672671-70672671 on Build GRCh38
rs1038778257 Chr.2: 70672681-70672681 on Build GRCh38
rs1051765383 Chr.2: 70672744-70672744 on Build GRCh38
rs782391328 Chr.2: 70672859-70672859 on Build GRCh38
rs182891869 Chr.2: 70672800-70672800 on Build GRCh38
rs782410218 Chr.2: 70672880-70672880 on Build GRCh38
rs782629299 Chr.2: 70672858-70672858 on Build GRCh38
rs781982849 Chr.2: 70672868-70672868 on Build GRCh38
rs533584493 Chr.2: 70672807-70672807 on Build GRCh38
rs781937169 Chr.2: 70672705-70672706 on Build GRCh38
rs933030954 Chr.2: 70672717-70672717 on Build GRCh38
rs782173852 Chr.2: 70672878-70672878 on Build GRCh38
rs771550887 Chr.2: 70672805-70672805 on Build GRCh38
rs922901507 Chr.2: 70672711-70672711 on Build GRCh38
rs782504722 Chr.2: 70672829-70672829 on Build GRCh38
rs202114130 Chr.2: 70672837-70672837 on Build GRCh38
rs1054594611 Chr.2: 70672811-70672811 on Build GRCh38
rs782682210 Chr.2: 70672830-70672830 on Build GRCh38
rs782441976 Chr.2: 70672771-70672771 on Build GRCh38

View Full Product Details