Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12350407-12350666 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF442
Entrez Gene ID
79973
Gene Name
zinc finger protein 442
Gene Aliases
-
UniGene
Hs.253193
Gene Chromosome Location
Chr.19: 12349371-12372636 on Build GRCh38
Transcripts
NM_030824.2 XM_006722908.3 XM_017027316.1 XM_017027317.1 XM_017027318.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4750091 c.1126C>A p.P376T Chr.19: 12350459-12350459 on Build GRCh38
265019 c.1106G>T p.R369I Chr.19: 12350479-12350479 on Build GRCh38
4750092 c.1084A>G p.S362G Chr.19: 12350501-12350501 on Build GRCh38
1234066 c.1004G>A p.S335N Chr.19: 12350581-12350581 on Build GRCh38
383938 c.1125A>C p.K375N Chr.19: 12350460-12350460 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs907201577 Chr.19: 12350442-12350442 on Build GRCh38
rs774736416 Chr.19: 12350527-12350527 on Build GRCh38
rs777866709 Chr.19: 12350488-12350488 on Build GRCh38
rs752881928 Chr.19: 12350487-12350487 on Build GRCh38
rs200426303 Chr.19: 12350590-12350590 on Build GRCh38
rs371288866 Chr.19: 12350529-12350529 on Build GRCh38
rs776342454 Chr.19: 12350517-12350517 on Build GRCh38
rs778862303 Chr.19: 12350606-12350606 on Build GRCh38
rs753016008 Chr.19: 12350445-12350446 on Build GRCh38
rs144013357 Chr.19: 12350607-12350607 on Build GRCh38
rs374264593 Chr.19: 12350598-12350598 on Build GRCh38
rs758906314 Chr.19: 12350515-12350516 on Build GRCh38
rs201256862 Chr.19: 12350463-12350464 on Build GRCh38
rs758182364 Chr.19: 12350616-12350616 on Build GRCh38
rs755304276 Chr.19: 12350471-12350471 on Build GRCh38
rs763553340 Chr.19: 12350520-12350520 on Build GRCh38
rs768259957 Chr.19: 12350516-12350516 on Build GRCh38
rs779764337 Chr.19: 12350623-12350623 on Build GRCh38
rs1020883043 Chr.19: 12350549-12350549 on Build GRCh38
rs750928971 Chr.19: 12350432-12350432 on Build GRCh38
rs771787039 Chr.19: 12350498-12350498 on Build GRCh38
rs766845359 Chr.19: 12350525-12350525 on Build GRCh38
rs766840840 Chr.19: 12350451-12350451 on Build GRCh38
rs967535082 Chr.19: 12350554-12350554 on Build GRCh38
rs746650695 Chr.19: 12350628-12350628 on Build GRCh38
rs551604101 Chr.19: 12350640-12350641 on Build GRCh38
rs749201526 Chr.19: 12350491-12350491 on Build GRCh38
rs752031955 Chr.19: 12350458-12350458 on Build GRCh38
rs781415922 Chr.19: 12350479-12350479 on Build GRCh38
rs764145174 Chr.19: 12350593-12350593 on Build GRCh38
rs149099428 Chr.19: 12350493-12350493 on Build GRCh38
rs1001569831 Chr.19: 12350443-12350443 on Build GRCh38
rs757332272 Chr.19: 12350600-12350600 on Build GRCh38
rs746699758 Chr.19: 12350504-12350504 on Build GRCh38
rs745608663 Chr.19: 12350497-12350497 on Build GRCh38
rs763607814 Chr.19: 12350448-12350448 on Build GRCh38
rs778567234 Chr.19: 12350496-12350496 on Build GRCh38
rs191332227 Chr.19: 12350431-12350431 on Build GRCh38
rs572669414 Chr.19: 12350502-12350502 on Build GRCh38
rs201428588 Chr.19: 12350577-12350577 on Build GRCh38
rs767800692 Chr.19: 12350562-12350562 on Build GRCh38

View Full Product Details