Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 40532516-40532779 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF621
Entrez Gene ID
285268
Gene Name
zinc finger protein 621
Gene Aliases
-
UniGene
Hs.19977
Gene Chromosome Location
Chr.3: 40523854-40539794 on Build GRCh38
Transcripts
NM_001098414.2 NM_001287245.1 NM_198484.4 XM_005265079.3 XM_005265080.3 XM_006713114.3 XM_011533610.2 XM_017006226.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
278376 c.932G>T p.R311I Chr.3: 40532702-40532702 on Build GRCh38
278377 c.1001T>G p.F334C Chr.3: 40532771-40532771 on Build GRCh38
1137026 c.873T>A p.Y291* Chr.3: 40532643-40532643 on Build GRCh38
7129392 c.778G>C p.E260Q Chr.3: 40532548-40532548 on Build GRCh38
1234370 c.853C>A p.H285N Chr.3: 40532623-40532623 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750147067 Chr.3: 40532699-40532700 on Build GRCh38
rs566518778 Chr.3: 40532586-40532586 on Build GRCh38
rs376957315 Chr.3: 40532543-40532543 on Build GRCh38
rs764297670 Chr.3: 40532591-40532591 on Build GRCh38
rs772378254 Chr.3: 40532735-40532735 on Build GRCh38
rs770923083 Chr.3: 40532585-40532585 on Build GRCh38
rs376371090 Chr.3: 40532694-40532694 on Build GRCh38
rs762076186 Chr.3: 40532612-40532612 on Build GRCh38
rs536118418 Chr.3: 40532573-40532573 on Build GRCh38
rs780369543 Chr.3: 40532627-40532627 on Build GRCh38
rs754442540 Chr.3: 40532648-40532648 on Build GRCh38
rs61740750 Chr.3: 40532580-40532580 on Build GRCh38
rs1036122937 Chr.3: 40532738-40532738 on Build GRCh38
rs200535212 Chr.3: 40532685-40532685 on Build GRCh38
rs773438210 Chr.3: 40532737-40532737 on Build GRCh38
rs141557940 Chr.3: 40532548-40532548 on Build GRCh38
rs750749480 Chr.3: 40532618-40532618 on Build GRCh38
rs765379156 Chr.3: 40532731-40532731 on Build GRCh38
rs776378551 Chr.3: 40532617-40532617 on Build GRCh38
rs761914937 Chr.3: 40532727-40532727 on Build GRCh38
rs373080077 Chr.3: 40532605-40532605 on Build GRCh38
rs866215627 Chr.3: 40532667-40532667 on Build GRCh38
rs537594378 Chr.3: 40532712-40532712 on Build GRCh38
rs538804516 Chr.3: 40532587-40532587 on Build GRCh38
rs765174745 Chr.3: 40532748-40532748 on Build GRCh38
rs202067818 Chr.3: 40532683-40532683 on Build GRCh38
rs574983572 Chr.3: 40532711-40532711 on Build GRCh38
rs940456661 Chr.3: 40532728-40532728 on Build GRCh38
rs911648421 Chr.3: 40532630-40532630 on Build GRCh38
rs777571744 Chr.3: 40532672-40532672 on Build GRCh38
rs140445368 Chr.3: 40532703-40532703 on Build GRCh38
rs752166883 Chr.3: 40532643-40532643 on Build GRCh38
rs774146837 Chr.3: 40532698-40532698 on Build GRCh38
rs758767315 Chr.3: 40532624-40532624 on Build GRCh38
rs769688766 Chr.3: 40532574-40532574 on Build GRCh38
rs867798760 Chr.3: 40532704-40532704 on Build GRCh38
rs370077146 Chr.3: 40532544-40532544 on Build GRCh38
rs747633470 Chr.3: 40532655-40532655 on Build GRCh38
rs765469170 Chr.3: 40532616-40532616 on Build GRCh38
rs768227630 Chr.3: 40532556-40532556 on Build GRCh38
rs767551214 Chr.3: 40532558-40532558 on Build GRCh38
rs138317598 Chr.3: 40532652-40532652 on Build GRCh38
rs957388992 Chr.3: 40532554-40532555 on Build GRCh38
rs989726252 Chr.3: 40532674-40532674 on Build GRCh38

View Full Product Details