Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 2885072-2885321 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDKN1C
Entrez Gene ID
1028
Gene Name
cyclin dependent kinase inhibitor 1C
Gene Aliases
BWCR, BWS, KIP2, WBS, p57, p57Kip2
UniGene
Hs.106070
Gene Chromosome Location
Chr.11: 2883218-2885804 on Build GRCh38
Transcripts
NM_000076.2 NM_001122630.1 NM_001122631.1 XM_005252732.4 XM_017017088.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6376658 c.232C>T p.Q78* Chr.11: 2885258-2885258 on Build GRCh38
4939514 c.283G>A p.V95M Chr.11: 2885207-2885207 on Build GRCh38
1178892 c.196delC p.Q66fs*206 Chr.11: 2885294-2885294 on Build GRCh38
4748798 c.262C>T p.P88S Chr.11: 2885228-2885228 on Build GRCh38
3943620 c.303C>T p.R101R Chr.11: 2885187-2885187 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750526402 Chr.11: 2885264-2885264 on Build GRCh38
rs764328739 Chr.11: 2885181-2885181 on Build GRCh38
rs483352974 Chr.11: 2885182-2885183 on Build GRCh38
rs931922290 Chr.11: 2885154-2885154 on Build GRCh38
rs534471786 Chr.11: 2885166-2885166 on Build GRCh38
rs483352971 Chr.11: 2885272-2885272 on Build GRCh38
rs1038466183 Chr.11: 2885249-2885249 on Build GRCh38
rs774805080 Chr.11: 2885185-2885185 on Build GRCh38
rs1008606799 Chr.11: 2885226-2885226 on Build GRCh38
rs932963247 Chr.11: 2885163-2885163 on Build GRCh38
rs747757242 Chr.11: 2885127-2885127 on Build GRCh38
rs483352970 Chr.11: 2885281-2885281 on Build GRCh38
rs483352972 Chr.11: 2885235-2885236 on Build GRCh38
rs786205235 Chr.11: 2885156-2885157 on Build GRCh38
rs763525000 Chr.11: 2885170-2885170 on Build GRCh38
rs897964106 Chr.11: 2885233-2885233 on Build GRCh38
rs778520280 Chr.11: 2885303-2885303 on Build GRCh38
rs570636789 Chr.11: 2885141-2885141 on Build GRCh38
rs141005361 Chr.11: 2885244-2885244 on Build GRCh38
rs551863674 Chr.11: 2885126-2885126 on Build GRCh38
rs753651871 Chr.11: 2885282-2885282 on Build GRCh38
rs387906399 Chr.11: 2885179-2885180 on Build GRCh38
rs772684721 Chr.11: 2885138-2885138 on Build GRCh38
rs930266866 Chr.11: 2885293-2885293 on Build GRCh38
rs483352975 Chr.11: 2885169-2885169 on Build GRCh38
rs483352969 Chr.11: 2885298-2885300 on Build GRCh38
rs1015104953 Chr.11: 2885232-2885232 on Build GRCh38
rs778772076 Chr.11: 2885100-2885100 on Build GRCh38
rs483352973 Chr.11: 2885225-2885226 on Build GRCh38
rs400377 Chr.11: 2885210-2885210 on Build GRCh38
rs762106424 Chr.11: 2885207-2885207 on Build GRCh38
rs771731330 Chr.11: 2885137-2885137 on Build GRCh38
rs945890937 Chr.11: 2885140-2885140 on Build GRCh38
rs1013695050 Chr.11: 2885090-2885090 on Build GRCh38
rs756321814 Chr.11: 2885271-2885271 on Build GRCh38
rs904430375 Chr.11: 2885151-2885151 on Build GRCh38
rs770277810 Chr.11: 2885145-2885145 on Build GRCh38
rs923004197 Chr.11: 2885160-2885160 on Build GRCh38
rs754972138 Chr.11: 2885301-2885301 on Build GRCh38

View Full Product Details