Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 173885600-173885845 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZBTB37
Entrez Gene ID
84614
Gene Name
zinc finger and BTB domain containing 37
Gene Aliases
D430004I08Rik, ZNF908
UniGene
Hs.668497
Gene Chromosome Location
Chr.1: 173868095-173891122 on Build GRCh38
Transcripts
NM_001122770.1 XM_005245546.2 XM_006711578.3 XM_011510062.2 XM_017002556.1 XM_017002557.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4459119 c.1112C>T p.P371L Chr.1: 173885724-173885724 on Build GRCh38
1336373 c.1187_1188insG p.I398fs*70 Chr.1: 173885799-173885800 on Build GRCh38
2075800 c.1102C>T p.R368W Chr.1: 173885714-173885714 on Build GRCh38
6541256 c.1044G>T p.M348I Chr.1: 173885656-173885656 on Build GRCh38
4839210 c.1202T>C p.F401S Chr.1: 173885814-173885814 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762488647 Chr.1: 173885747-173885747 on Build GRCh38
rs945743529 Chr.1: 173885655-173885655 on Build GRCh38
rs987302525 Chr.1: 173885746-173885746 on Build GRCh38
rs766985897 Chr.1: 173885683-173885683 on Build GRCh38
rs925586490 Chr.1: 173885706-173885706 on Build GRCh38
rs752272147 Chr.1: 173885715-173885715 on Build GRCh38
rs773686041 Chr.1: 173885667-173885667 on Build GRCh38
rs963601669 Chr.1: 173885671-173885671 on Build GRCh38
rs971936499 Chr.1: 173885681-173885681 on Build GRCh38
rs373338183 Chr.1: 173885679-173885679 on Build GRCh38
rs916995618 Chr.1: 173885638-173885638 on Build GRCh38
rs1054431073 Chr.1: 173885724-173885724 on Build GRCh38
rs935693688 Chr.1: 173885718-173885718 on Build GRCh38
rs1041229698 Chr.1: 173885822-173885822 on Build GRCh38
rs763842910 Chr.1: 173885770-173885770 on Build GRCh38
rs761301315 Chr.1: 173885670-173885670 on Build GRCh38
rs188075755 Chr.1: 173885782-173885782 on Build GRCh38
rs1041469753 Chr.1: 173885660-173885660 on Build GRCh38

View Full Product Details