Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 101651248-101651499 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZBTB11
Entrez Gene ID
27107
Gene Name
zinc finger and BTB domain containing 11
Gene Aliases
ZNF-U69274, ZNF913
UniGene
Hs.655286
Gene Chromosome Location
Chr.3: 101649439-101677144 on Build GRCh38
Transcripts
NM_014415.3 XM_011512689.2 XM_011512690.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6095445 c.3072A>T p.L1024F Chr.3: 101651256-101651256 on Build GRCh38
3766823 c.3041A>G p.Q1014R Chr.3: 101651287-101651287 on Build GRCh38
6483002 c.2992G>A p.A998T Chr.3: 101651336-101651336 on Build GRCh38
7316969 c.3035C>T p.S1012F Chr.3: 101651293-101651293 on Build GRCh38
6498387 c.2857C>T p.Q953* Chr.3: 101651471-101651471 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368255825 Chr.3: 101651451-101651451 on Build GRCh38
rs781611958 Chr.3: 101651378-101651378 on Build GRCh38
rs773075411 Chr.3: 101651287-101651287 on Build GRCh38
rs1010944646 Chr.3: 101651427-101651427 on Build GRCh38
rs147439598 Chr.3: 101651324-101651324 on Build GRCh38
rs753198699 Chr.3: 101651341-101651341 on Build GRCh38
rs764439208 Chr.3: 101651351-101651351 on Build GRCh38
rs769268626 Chr.3: 101651452-101651452 on Build GRCh38
rs199696195 Chr.3: 101651307-101651307 on Build GRCh38
rs374150569 Chr.3: 101651403-101651403 on Build GRCh38
rs549056576 Chr.3: 101651414-101651414 on Build GRCh38
rs775066854 Chr.3: 101651458-101651458 on Build GRCh38
rs755795557 Chr.3: 101651319-101651320 on Build GRCh38
rs752000475 Chr.3: 101651367-101651367 on Build GRCh38
rs80262325 Chr.3: 101651376-101651376 on Build GRCh38
rs138181654 Chr.3: 101651388-101651388 on Build GRCh38
rs762326730 Chr.3: 101651460-101651460 on Build GRCh38
rs569562978 Chr.3: 101651331-101651331 on Build GRCh38
rs1054519351 Chr.3: 101651396-101651396 on Build GRCh38
rs150207556 Chr.3: 101651308-101651308 on Build GRCh38
rs780169867 Chr.3: 101651408-101651408 on Build GRCh38
rs566978820 Chr.3: 101651445-101651445 on Build GRCh38
rs750642606 Chr.3: 101651400-101651400 on Build GRCh38
rs776310585 Chr.3: 101651448-101651448 on Build GRCh38
rs777533902 Chr.3: 101651422-101651422 on Build GRCh38
rs964275700 Chr.3: 101651387-101651387 on Build GRCh38
rs1041966107 Chr.3: 101651432-101651432 on Build GRCh38
rs1031052442 Chr.3: 101651304-101651304 on Build GRCh38
rs770692954 Chr.3: 101651447-101651447 on Build GRCh38
rs955733469 Chr.3: 101651348-101651348 on Build GRCh38
rs760396359 Chr.3: 101651295-101651295 on Build GRCh38
rs749270421 Chr.3: 101651412-101651412 on Build GRCh38
rs771821359 Chr.3: 101651284-101651284 on Build GRCh38
rs748112807 Chr.3: 101651281-101651281 on Build GRCh38
rs920249837 Chr.3: 101651441-101651441 on Build GRCh38
rs370880069 Chr.3: 101651342-101651342 on Build GRCh38

View Full Product Details