Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 57456121-57456352 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VN1R1
Entrez Gene ID
57191
Gene Name
vomeronasal 1 receptor 1
Gene Aliases
V1RL1, VNR19I1, ZVNH1, ZVNR1
UniGene
Hs.677423
Gene Chromosome Location
Chr.19: 57455174-57456739 on Build GRCh38
Transcripts
NM_020633.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1397046 c.209A>T p.Y70F Chr.19: 57456278-57456278 on Build GRCh38
7115496 c.165G>C p.Q55H Chr.19: 57456322-57456322 on Build GRCh38
7098996 c.313C>G p.Q105E Chr.19: 57456174-57456174 on Build GRCh38
1397045 c.248C>T p.T83M Chr.19: 57456239-57456239 on Build GRCh38
6839980 c.268C>G p.L90V Chr.19: 57456219-57456219 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200564544 Chr.19: 57456291-57456291 on Build GRCh38
rs780239197 Chr.19: 57456188-57456188 on Build GRCh38
rs372495902 Chr.19: 57456239-57456239 on Build GRCh38
rs762326806 Chr.19: 57456254-57456254 on Build GRCh38
rs779031970 Chr.19: 57456320-57456320 on Build GRCh38
rs754966658 Chr.19: 57456305-57456305 on Build GRCh38
rs139353138 Chr.19: 57456233-57456233 on Build GRCh38
rs749270907 Chr.19: 57456210-57456210 on Build GRCh38
rs376448405 Chr.19: 57456289-57456289 on Build GRCh38
rs757567592 Chr.19: 57456167-57456167 on Build GRCh38
rs768163764 Chr.19: 57456228-57456228 on Build GRCh38
rs147292022 Chr.19: 57456157-57456157 on Build GRCh38
rs200574657 Chr.19: 57456272-57456273 on Build GRCh38
rs759601493 Chr.19: 57456235-57456235 on Build GRCh38
rs144835279 Chr.19: 57456270-57456270 on Build GRCh38
rs776929276 Chr.19: 57456318-57456319 on Build GRCh38
rs768928914 Chr.19: 57456220-57456220 on Build GRCh38
rs114002003 Chr.19: 57456295-57456295 on Build GRCh38
rs756609780 Chr.19: 57456183-57456183 on Build GRCh38
rs748424838 Chr.19: 57456321-57456321 on Build GRCh38
rs751261130 Chr.19: 57456257-57456257 on Build GRCh38
rs900497265 Chr.19: 57456296-57456296 on Build GRCh38
rs755505752 Chr.19: 57456200-57456200 on Build GRCh38
rs781514672 Chr.19: 57456177-57456177 on Build GRCh38
rs990648071 Chr.19: 57456222-57456222 on Build GRCh38
rs756628244 Chr.19: 57456276-57456276 on Build GRCh38
rs553434064 Chr.19: 57456223-57456223 on Build GRCh38
rs143275734 Chr.19: 57456238-57456238 on Build GRCh38
rs763828507 Chr.19: 57456256-57456256 on Build GRCh38
rs746824707 Chr.19: 57456225-57456225 on Build GRCh38
rs750773883 Chr.19: 57456271-57456271 on Build GRCh38
rs756441436 Chr.19: 57456280-57456280 on Build GRCh38
rs3746223 Chr.19: 57456178-57456178 on Build GRCh38
rs867865910 Chr.19: 57456203-57456203 on Build GRCh38

View Full Product Details