Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 8838271-8838506 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
A2ML1
Entrez Gene ID
144568
Gene Name
alpha-2-macroglobulin like 1
Gene Aliases
CPAMD9
UniGene
Hs.620532
Gene Chromosome Location
Chr.12: 8822472-8887202 on Build GRCh38
Transcripts
NM_144670.5 XM_011520566.2 XM_011520567.2 XM_017018868.1 XM_017018869.1 XM_017018870.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3753524 c.861C>A p.D287E Chr.12: 8838341-8838341 on Build GRCh38
4657808 c.865A>G p.T289A Chr.12: 8838345-8838345 on Build GRCh38
268365 c.900C>A p.T300T Chr.12: 8838380-8838380 on Build GRCh38
3466262 c.907C>G p.L303V Chr.12: 8838387-8838387 on Build GRCh38
5013939 c.856-2A>T p.? Chr.12: 8838334-8838334 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773198059 Chr.12: 8838467-8838467 on Build GRCh38
rs776730430 Chr.12: 8838459-8838459 on Build GRCh38
rs889804273 Chr.12: 8838421-8838421 on Build GRCh38
rs778541050 Chr.12: 8838440-8838440 on Build GRCh38
rs184575465 Chr.12: 8838391-8838391 on Build GRCh38
rs746280503 Chr.12: 8838436-8838436 on Build GRCh38
rs779544502 Chr.12: 8838427-8838427 on Build GRCh38
rs751287968 Chr.12: 8838444-8838444 on Build GRCh38
rs61921916 Chr.12: 8838341-8838341 on Build GRCh38
rs769781211 Chr.12: 8838466-8838466 on Build GRCh38
rs762795796 Chr.12: 8838393-8838393 on Build GRCh38
rs199621515 Chr.12: 8838368-8838368 on Build GRCh38
rs746717919 Chr.12: 8838485-8838488 on Build GRCh38
rs896631846 Chr.12: 8838484-8838484 on Build GRCh38
rs779735317 Chr.12: 8838312-8838312 on Build GRCh38
rs117140170 Chr.12: 8838317-8838317 on Build GRCh38
rs765372447 Chr.12: 8838404-8838404 on Build GRCh38
rs192888493 Chr.12: 8838367-8838367 on Build GRCh38
rs372520121 Chr.12: 8838401-8838401 on Build GRCh38
rs763486898 Chr.12: 8838423-8838423 on Build GRCh38
rs779931920 Chr.12: 8838481-8838482 on Build GRCh38
rs189848036 Chr.12: 8838400-8838400 on Build GRCh38
rs757793092 Chr.12: 8838334-8838334 on Build GRCh38
rs765127537 Chr.12: 8838384-8838384 on Build GRCh38
rs758026611 Chr.12: 8838310-8838310 on Build GRCh38
rs749829095 Chr.12: 8838332-8838332 on Build GRCh38
rs75617691 Chr.12: 8838343-8838343 on Build GRCh38
rs768595376 Chr.12: 8838484-8838485 on Build GRCh38
rs775772721 Chr.12: 8838361-8838361 on Build GRCh38
rs762733139 Chr.12: 8838473-8838473 on Build GRCh38
rs761769559 Chr.12: 8838380-8838380 on Build GRCh38
rs1054572824 Chr.12: 8838460-8838460 on Build GRCh38
rs771580168 Chr.12: 8838379-8838379 on Build GRCh38
rs752217277 Chr.12: 8838414-8838414 on Build GRCh38
rs757883065 Chr.12: 8838422-8838422 on Build GRCh38
rs754507474 Chr.12: 8838321-8838321 on Build GRCh38
rs766211795 Chr.12: 8838480-8838480 on Build GRCh38
rs768824265 Chr.12: 8838454-8838454 on Build GRCh38
rs780222495 Chr.12: 8838446-8838446 on Build GRCh38
rs748203389 Chr.12: 8838463-8838463 on Build GRCh38
rs780784023 Chr.12: 8838326-8838326 on Build GRCh38
rs61745855 Chr.12: 8838447-8838447 on Build GRCh38
rs776712868 Chr.12: 8838347-8838347 on Build GRCh38
rs750178152 Chr.12: 8838307-8838307 on Build GRCh38
rs776642180 Chr.12: 8838372-8838372 on Build GRCh38

View Full Product Details