Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 70816878-70817143 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC4F
Entrez Gene ID
165530
Gene Name
C-type lectin domain family 4 member F
Gene Aliases
CLECSF13, KCLR
UniGene
Hs.681239
Gene Chromosome Location
Chr.2: 70808643-70825235 on Build GRCh38
Transcripts
NM_001258027.1 NM_001321308.1 NM_173535.2 XM_011532635.2 XM_011532637.2 XM_011532638.2 XM_011532639.2 XM_011532640.2 XM_011532641.2 XM_011532642.2 XM_011532643.1 XM_017003519.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5862774 c.400C>T p.Q134* Chr.2: 70816981-70816981 on Build GRCh38
395244 c.454G>T p.V152L Chr.2: 70816927-70816927 on Build GRCh38
1201360 c.431A>G p.N144S Chr.2: 70816950-70816950 on Build GRCh38
4803736 c.450G>A p.Q150Q Chr.2: 70816931-70816931 on Build GRCh38
4394077 c.405C>T p.L135L Chr.2: 70816976-70816976 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782073242 Chr.2: 70816925-70816925 on Build GRCh38
rs368099509 Chr.2: 70817108-70817108 on Build GRCh38
rs782035331 Chr.2: 70817100-70817100 on Build GRCh38
rs782324435 Chr.2: 70817110-70817110 on Build GRCh38
rs781787197 Chr.2: 70817035-70817035 on Build GRCh38
rs781927762 Chr.2: 70816913-70816913 on Build GRCh38
rs540670274 Chr.2: 70817018-70817018 on Build GRCh38
rs146659719 Chr.2: 70817025-70817025 on Build GRCh38
rs782567216 Chr.2: 70816966-70816966 on Build GRCh38
rs781805654 Chr.2: 70816967-70816967 on Build GRCh38
rs61754946 Chr.2: 70816944-70816944 on Build GRCh38
rs377254965 Chr.2: 70817011-70817011 on Build GRCh38
rs546185298 Chr.2: 70817080-70817080 on Build GRCh38
rs782182562 Chr.2: 70816987-70816987 on Build GRCh38
rs782330890 Chr.2: 70817001-70817001 on Build GRCh38
rs781885536 Chr.2: 70816958-70816958 on Build GRCh38
rs116477018 Chr.2: 70817008-70817008 on Build GRCh38
rs972365529 Chr.2: 70816910-70816910 on Build GRCh38
rs368623325 Chr.2: 70817064-70817064 on Build GRCh38
rs781833119 Chr.2: 70817044-70817044 on Build GRCh38
rs913879369 Chr.2: 70817007-70817007 on Build GRCh38
rs782793654 Chr.2: 70816953-70816953 on Build GRCh38
rs555202980 Chr.2: 70816983-70816983 on Build GRCh38
rs117307369 Chr.2: 70816929-70816929 on Build GRCh38
rs920830151 Chr.2: 70816911-70816911 on Build GRCh38
rs183733688 Chr.2: 70816982-70816982 on Build GRCh38
rs563171601 Chr.2: 70817081-70817081 on Build GRCh38
rs903765256 Chr.2: 70817032-70817032 on Build GRCh38
rs782698487 Chr.2: 70816984-70816984 on Build GRCh38
rs200228677 Chr.2: 70817116-70817116 on Build GRCh38
rs782724014 Chr.2: 70817037-70817037 on Build GRCh38
rs782000818 Chr.2: 70817026-70817026 on Build GRCh38
rs782735011 Chr.2: 70816928-70816928 on Build GRCh38
rs782468347 Chr.2: 70816976-70816976 on Build GRCh38
rs952245427 Chr.2: 70816924-70816924 on Build GRCh38
rs782265741 Chr.2: 70817091-70817091 on Build GRCh38
rs782562166 Chr.2: 70816963-70816963 on Build GRCh38
rs781908895 Chr.2: 70817069-70817069 on Build GRCh38
rs944758850 Chr.2: 70816946-70816946 on Build GRCh38
rs2075221 Chr.2: 70817079-70817079 on Build GRCh38
rs141757870 Chr.2: 70816931-70816931 on Build GRCh38
rs782498429 Chr.2: 70817053-70817053 on Build GRCh38
rs375483366 Chr.2: 70817098-70817098 on Build GRCh38

View Full Product Details