Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 73385868-73386141 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALMS1
Entrez Gene ID
7840
Gene Name
ALMS1, centrosome and basal body associated protein
Gene Aliases
ALSS
UniGene
Hs.184720
Gene Chromosome Location
Chr.2: 73385758-73609919 on Build GRCh38
Transcripts
NM_015120.4

SNP Detail

SNP ID Chromosome Location
rs906421049 Chr.2: 73385920-73385920 on Build GRCh38
rs759871854 Chr.2: 73386069-73386069 on Build GRCh38
rs1006328829 Chr.2: 73385935-73385936 on Build GRCh38
rs1045405319 Chr.2: 73385896-73385896 on Build GRCh38
rs777844728 Chr.2: 73385932-73385933 on Build GRCh38
rs70965731 Chr.2: 73385940-73385942 on Build GRCh38
rs1054040147 Chr.2: 73385937-73385937 on Build GRCh38
rs61156725 Chr.2: 73385937-73385942 on Build GRCh38
rs767731236 Chr.2: 73385982-73385982 on Build GRCh38
rs1025593035 Chr.2: 73385956-73385956 on Build GRCh38
rs797045229 Chr.2: 73385945-73385953 on Build GRCh38
rs780977235 Chr.2: 73385959-73385964 on Build GRCh38
rs866503570 Chr.2: 73385925-73385925 on Build GRCh38
rs745737550 Chr.2: 73386028-73386028 on Build GRCh38
rs765378599 Chr.2: 73386071-73386071 on Build GRCh38
rs766851353 Chr.2: 73386059-73386059 on Build GRCh38
rs762635802 Chr.2: 73386086-73386087 on Build GRCh38
rs765418565 Chr.2: 73386003-73386003 on Build GRCh38
rs571417117 Chr.2: 73386017-73386017 on Build GRCh38
rs756301830 Chr.2: 73385931-73385932 on Build GRCh38
rs199611703 Chr.2: 73385902-73385902 on Build GRCh38
rs779179743 Chr.2: 73385943-73385944 on Build GRCh38
rs778264668 Chr.2: 73386109-73386109 on Build GRCh38
rs772450464 Chr.2: 73386117-73386117 on Build GRCh38
rs753006733 Chr.2: 73385928-73385928 on Build GRCh38
rs762403314 Chr.2: 73385978-73385978 on Build GRCh38
rs778948871 Chr.2: 73385909-73385910 on Build GRCh38
rs772724963 Chr.2: 73385904-73385906 on Build GRCh38
rs1016245881 Chr.2: 73385948-73385948 on Build GRCh38
rs755760721 Chr.2: 73386029-73386029 on Build GRCh38
rs537537228 Chr.2: 73385988-73385988 on Build GRCh38
rs864622738 Chr.2: 73385934-73385942 on Build GRCh38
rs762436798 Chr.2: 73385904-73385915 on Build GRCh38
rs769538172 Chr.2: 73385893-73385893 on Build GRCh38
rs748863552 Chr.2: 73385955-73385955 on Build GRCh38
rs13009604 Chr.2: 73385946-73385946 on Build GRCh38
rs866402031 Chr.2: 73386095-73386095 on Build GRCh38
rs368947491 Chr.2: 73386102-73386102 on Build GRCh38
rs55889738 Chr.2: 73385942-73385943 on Build GRCh38
rs973057702 Chr.2: 73385956-73385958 on Build GRCh38
rs755038994 Chr.2: 73386001-73386001 on Build GRCh38
rs757262099 Chr.2: 73386018-73386018 on Build GRCh38
rs767882166 Chr.2: 73385925-73385926 on Build GRCh38
rs767558791 Chr.2: 73386086-73386086 on Build GRCh38
rs886056296 Chr.2: 73385970-73385971 on Build GRCh38
rs905301935 Chr.2: 73385929-73385930 on Build GRCh38
rs1037793096 Chr.2: 73386101-73386101 on Build GRCh38
rs894068220 Chr.2: 73386005-73386005 on Build GRCh38
rs878854999 Chr.2: 73385925-73385942 on Build GRCh38
rs774968380 Chr.2: 73385954-73385954 on Build GRCh38
rs747912991 Chr.2: 73385994-73385995 on Build GRCh38
rs201868751 Chr.2: 73385989-73385989 on Build GRCh38
rs373614102 Chr.2: 73385889-73385889 on Build GRCh38
rs955371491 Chr.2: 73385971-73385971 on Build GRCh38
rs754175473 Chr.2: 73385938-73385943 on Build GRCh38
rs905373886 Chr.2: 73386087-73386087 on Build GRCh38
rs747750620 Chr.2: 73386113-73386113 on Build GRCh38
rs755530002 Chr.2: 73385919-73385923 on Build GRCh38
rs774784126 Chr.2: 73385977-73385977 on Build GRCh38
rs778271633 Chr.2: 73386120-73386120 on Build GRCh38
rs193922695 Chr.2: 73385939-73385940 on Build GRCh38
rs755847221 Chr.2: 73386096-73386096 on Build GRCh38
rs772683919 Chr.2: 73386044-73386044 on Build GRCh38
rs753958280 Chr.2: 73386000-73386000 on Build GRCh38
rs768161233 Chr.2: 73385962-73385962 on Build GRCh38
rs757131983 Chr.2: 73386079-73386079 on Build GRCh38
rs761163393 Chr.2: 73386057-73386057 on Build GRCh38
rs754662930 Chr.2: 73386103-73386103 on Build GRCh38
rs751415466 Chr.2: 73386010-73386010 on Build GRCh38
rs773881728 Chr.2: 73386037-73386037 on Build GRCh38
rs746896173 Chr.2: 73385958-73385959 on Build GRCh38
rs748571359 Chr.2: 73386041-73386041 on Build GRCh38
rs878853007 Chr.2: 73385907-73385909 on Build GRCh38
rs752874408 Chr.2: 73386075-73386075 on Build GRCh38
rs748955296 Chr.2: 73386036-73386036 on Build GRCh38
rs13009609 Chr.2: 73385961-73385961 on Build GRCh38
rs764804760 Chr.2: 73385903-73385904 on Build GRCh38
rs549739737 Chr.2: 73386016-73386016 on Build GRCh38
rs879255406 Chr.2: 73385937-73385939 on Build GRCh38
rs752032517 Chr.2: 73385916-73385916 on Build GRCh38
rs939208094 Chr.2: 73385923-73385923 on Build GRCh38
rs1004961829 Chr.2: 73385931-73385931 on Build GRCh38
rs773431417 Chr.2: 73386049-73386049 on Build GRCh38
rs200709146 Chr.2: 73385903-73385903 on Build GRCh38
rs766762252 Chr.2: 73385994-73385994 on Build GRCh38
rs1025964615 Chr.2: 73386119-73386119 on Build GRCh38
rs13009043 Chr.2: 73385943-73385943 on Build GRCh38
rs878855000 Chr.2: 73385928-73385942 on Build GRCh38
rs779615407 Chr.2: 73386033-73386033 on Build GRCh38
rs772848105 Chr.2: 73386075-73386076 on Build GRCh38
rs747529471 Chr.2: 73386121-73386121 on Build GRCh38
rs780831604 Chr.2: 73386026-73386026 on Build GRCh38
rs769689052 Chr.2: 73386056-73386056 on Build GRCh38
rs183407241 Chr.2: 73385928-73385928 on Build GRCh38
rs903704799 Chr.2: 73386005-73386006 on Build GRCh38
rs386390463 Chr.2: 73385922-73385923 on Build GRCh38
rs750332908 Chr.2: 73386089-73386089 on Build GRCh38
rs776932244 Chr.2: 73386068-73386068 on Build GRCh38

View Full Product Details