Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32031439-32031712 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KHDRBS1
Entrez Gene ID
10657
Gene Name
KH RNA binding domain containing, signal transduction associated 1
Gene Aliases
Sam68, p62, p68
UniGene
Hs.445893
Gene Chromosome Location
Chr.1: 32013694-32060859 on Build GRCh38
Transcripts
NM_001271878.1 NM_006559.2 NR_073498.1 NR_073499.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1644702 c.512A>G p.N171S Chr.1: 32031528-32031528 on Build GRCh38
908226 c.529C>A p.L177I Chr.1: 32031545-32031545 on Build GRCh38
908227 c.552C>G p.I184M Chr.1: 32031568-32031568 on Build GRCh38
6126115 c.568G>C p.E190Q Chr.1: 32031584-32031584 on Build GRCh38
2077522 c.520G>T p.G174W Chr.1: 32031536-32031536 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369355024 Chr.1: 32031475-32031475 on Build GRCh38
rs761077941 Chr.1: 32031517-32031517 on Build GRCh38
rs200585423 Chr.1: 32031604-32031604 on Build GRCh38
rs373088925 Chr.1: 32031497-32031497 on Build GRCh38
rs761314139 Chr.1: 32031554-32031554 on Build GRCh38
rs775962905 Chr.1: 32031514-32031514 on Build GRCh38
rs376286662 Chr.1: 32031505-32031505 on Build GRCh38
rs753283735 Chr.1: 32031521-32031521 on Build GRCh38
rs778548188 Chr.1: 32031473-32031473 on Build GRCh38
rs771221823 Chr.1: 32031507-32031507 on Build GRCh38
rs758223817 Chr.1: 32031481-32031481 on Build GRCh38
rs759929273 Chr.1: 32031508-32031508 on Build GRCh38
rs779996884 Chr.1: 32031485-32031485 on Build GRCh38
rs746901910 Chr.1: 32031489-32031489 on Build GRCh38
rs376888111 Chr.1: 32031689-32031689 on Build GRCh38
rs772301856 Chr.1: 32031510-32031510 on Build GRCh38
rs779686927 Chr.1: 32031674-32031674 on Build GRCh38
rs373475108 Chr.1: 32031623-32031623 on Build GRCh38
rs952602016 Chr.1: 32031670-32031670 on Build GRCh38
rs764715808 Chr.1: 32031519-32031519 on Build GRCh38
rs764832191 Chr.1: 32031583-32031583 on Build GRCh38
rs1028200894 Chr.1: 32031667-32031667 on Build GRCh38
rs758099293 Chr.1: 32031650-32031650 on Build GRCh38
rs749339024 Chr.1: 32031690-32031690 on Build GRCh38
rs751439232 Chr.1: 32031685-32031685 on Build GRCh38
rs754859442 Chr.1: 32031687-32031687 on Build GRCh38
rs749438000 Chr.1: 32031506-32031506 on Build GRCh38

View Full Product Details