Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 184576857-184577100 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHB3
Entrez Gene ID
2049
Gene Name
EPH receptor B3
Gene Aliases
EK2, ETK2, HEK2, TYRO6
UniGene
Hs.2913
Gene Chromosome Location
Chr.3: 184561799-184582408 on Build GRCh38
Transcripts
NM_004443.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1753055 c.1151G>C p.G384A Chr.3: 184576980-184576980 on Build GRCh38
5995246 c.1030C>T p.R344* Chr.3: 184576859-184576859 on Build GRCh38
4856832 c.1249G>A p.A417T Chr.3: 184577078-184577078 on Build GRCh38
7158545 c.1160C>T p.S387L Chr.3: 184576989-184576989 on Build GRCh38
730190 c.1201C>A p.R401R Chr.3: 184577030-184577030 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201958581 Chr.3: 184576979-184576979 on Build GRCh38
rs755747537 Chr.3: 184577012-184577012 on Build GRCh38
rs746388630 Chr.3: 184576934-184576934 on Build GRCh38
rs763312420 Chr.3: 184576985-184576985 on Build GRCh38
rs773463961 Chr.3: 184576978-184576978 on Build GRCh38
rs781759967 Chr.3: 184576917-184576917 on Build GRCh38
rs752170254 Chr.3: 184576989-184576989 on Build GRCh38
rs770017111 Chr.3: 184576963-184576963 on Build GRCh38
rs140106380 Chr.3: 184577031-184577031 on Build GRCh38
rs777423504 Chr.3: 184577013-184577013 on Build GRCh38
rs533688795 Chr.3: 184577014-184577014 on Build GRCh38
rs535315602 Chr.3: 184577052-184577052 on Build GRCh38
rs981146326 Chr.3: 184576975-184576975 on Build GRCh38
rs748176396 Chr.3: 184577038-184577038 on Build GRCh38
rs145258842 Chr.3: 184577029-184577029 on Build GRCh38
rs757360039 Chr.3: 184576974-184576974 on Build GRCh38
rs141064214 Chr.3: 184577001-184577001 on Build GRCh38
rs775157731 Chr.3: 184577063-184577063 on Build GRCh38
rs947277288 Chr.3: 184577081-184577081 on Build GRCh38
rs566350665 Chr.3: 184577000-184577000 on Build GRCh38
rs760161735 Chr.3: 184577064-184577064 on Build GRCh38
rs760101021 Chr.3: 184576993-184576993 on Build GRCh38
rs773552328 Chr.3: 184577047-184577047 on Build GRCh38
rs867469671 Chr.3: 184576983-184576983 on Build GRCh38
rs200601511 Chr.3: 184576991-184576991 on Build GRCh38
rs752450110 Chr.3: 184576900-184576900 on Build GRCh38
rs549325655 Chr.3: 184576916-184576916 on Build GRCh38
rs773741899 Chr.3: 184577054-184577054 on Build GRCh38
rs768778880 Chr.3: 184576940-184576940 on Build GRCh38
rs752217008 Chr.3: 184577008-184577008 on Build GRCh38
rs777462615 Chr.3: 184576909-184576909 on Build GRCh38
rs144365078 Chr.3: 184576987-184576987 on Build GRCh38
rs762060127 Chr.3: 184576959-184576959 on Build GRCh38
rs776669177 Chr.3: 184576952-184576952 on Build GRCh38
rs567574380 Chr.3: 184577023-184577023 on Build GRCh38
rs375763872 Chr.3: 184577024-184577024 on Build GRCh38
rs1014568253 Chr.3: 184576922-184576922 on Build GRCh38
rs370364618 Chr.3: 184577051-184577051 on Build GRCh38
rs201322151 Chr.3: 184576901-184576901 on Build GRCh38
rs1023272361 Chr.3: 184576957-184576957 on Build GRCh38
rs768092104 Chr.3: 184577055-184577055 on Build GRCh38
rs144807386 Chr.3: 184577015-184577015 on Build GRCh38
rs767180194 Chr.3: 184576995-184576995 on Build GRCh38
rs778841600 Chr.3: 184576933-184576933 on Build GRCh38
rs962372314 Chr.3: 184576972-184576972 on Build GRCh38
rs769913740 Chr.3: 184577046-184577046 on Build GRCh38

View Full Product Details