Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 20354754-20355017 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PARP2
Entrez Gene ID
10038
Gene Name
poly(ADP-ribose) polymerase 2
Gene Aliases
ADPRT2, ADPRTL2, ADPRTL3, ARTD2, PARP-2, pADPRT-2
UniGene
Hs.409412
Gene Chromosome Location
Chr.14: 20343041-20357904 on Build GRCh38
Transcripts
NM_001042618.1 NM_005484.3 XM_005267247.3 XM_017020912.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3736094 c.740G>A p.R247Q Chr.14: 20354893-20354893 on Build GRCh38
469754 c.817G>A p.A273T Chr.14: 20354823-20354823 on Build GRCh38
5024521 c.690C>G p.Y230* Chr.14: 20354843-20354843 on Build GRCh38
4560683 c.705G>C p.K235N Chr.14: 20354858-20354858 on Build GRCh38
1640030 c.886C>T p.R296* Chr.14: 20354892-20354892 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752266352 Chr.14: 20354997-20354997 on Build GRCh38
rs778155437 Chr.14: 20354866-20354870 on Build GRCh38
rs1006715501 Chr.14: 20354885-20354885 on Build GRCh38
rs971373621 Chr.14: 20354933-20354933 on Build GRCh38
rs1034612223 Chr.14: 20354803-20354803 on Build GRCh38
rs781471821 Chr.14: 20354816-20354816 on Build GRCh38
rs3093925 Chr.14: 20354859-20354859 on Build GRCh38
rs371236543 Chr.14: 20354860-20354860 on Build GRCh38
rs918547481 Chr.14: 20354910-20354910 on Build GRCh38
rs747921516 Chr.14: 20354793-20354793 on Build GRCh38
rs755919869 Chr.14: 20354795-20354795 on Build GRCh38
rs371220029 Chr.14: 20354916-20354916 on Build GRCh38
rs774975124 Chr.14: 20354886-20354886 on Build GRCh38
rs143908458 Chr.14: 20354901-20354901 on Build GRCh38
rs776814629 Chr.14: 20354954-20354954 on Build GRCh38
rs201498500 Chr.14: 20354826-20354826 on Build GRCh38
rs762270539 Chr.14: 20354955-20354955 on Build GRCh38
rs771196478 Chr.14: 20354814-20354814 on Build GRCh38
rs749402496 Chr.14: 20354797-20354797 on Build GRCh38
rs375534938 Chr.14: 20354974-20354974 on Build GRCh38
rs769072188 Chr.14: 20354875-20354875 on Build GRCh38
rs3093927 Chr.14: 20354936-20354936 on Build GRCh38
rs199986271 Chr.14: 20354869-20354869 on Build GRCh38
rs772533365 Chr.14: 20354843-20354843 on Build GRCh38
rs140156644 Chr.14: 20354874-20354874 on Build GRCh38
rs902186193 Chr.14: 20354813-20354813 on Build GRCh38
rs781311187 Chr.14: 20354788-20354788 on Build GRCh38
rs774244546 Chr.14: 20354820-20354820 on Build GRCh38
rs941062257 Chr.14: 20354995-20354995 on Build GRCh38
rs1014294597 Chr.14: 20354853-20354855 on Build GRCh38
rs759112105 Chr.14: 20354985-20354985 on Build GRCh38
rs745974870 Chr.14: 20354829-20354829 on Build GRCh38
rs373691226 Chr.14: 20354934-20354934 on Build GRCh38
rs757379983 Chr.14: 20354922-20354922 on Build GRCh38
rs773702438 Chr.14: 20354989-20354989 on Build GRCh38
rs747453800 Chr.14: 20354950-20354950 on Build GRCh38
rs770460167 Chr.14: 20354966-20354966 on Build GRCh38
rs559797488 Chr.14: 20354914-20354914 on Build GRCh38
rs768972248 Chr.14: 20354870-20354870 on Build GRCh38
rs745922620 Chr.14: 20354935-20354935 on Build GRCh38
rs754915323 Chr.14: 20354782-20354782 on Build GRCh38
rs751074729 Chr.14: 20354892-20354892 on Build GRCh38
rs780538446 Chr.14: 20354947-20354947 on Build GRCh38
rs3093926 Chr.14: 20354893-20354893 on Build GRCh38
rs769223139 Chr.14: 20354953-20354953 on Build GRCh38
rs372225740 Chr.14: 20354956-20354956 on Build GRCh38
rs998064891 Chr.14: 20354819-20354819 on Build GRCh38

View Full Product Details