Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 24445072-24445345 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NIPAL3
Entrez Gene ID
57185
Gene Name
NIPA like domain containing 3
Gene Aliases
DJ462O23.2, NPAL3
UniGene
Hs.523442
Gene Chromosome Location
Chr.1: 24413360-24472983 on Build GRCh38
Transcripts
NM_001322854.1 NM_001322855.1 NM_001322856.1 NM_001322857.1 NM_001322858.1 NM_001322859.1 NM_001322860.1 NM_001322861.1 NM_001322862.1 NM_001322863.1 NM_001322864.1 NM_001322865.1 NM_001322866.1 NM_001322868.1 NM_020448.4 NR_136506.1 NR_136507.1 NR_136508.1 XM_011541808.2 XM_017001868.1 XM_017001869.1 XM_017001870.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6460314 c.371A>G p.K124R Chr.1: 24445221-24445221 on Build GRCh38
4833878 c.391C>T p.L131L Chr.1: 24445241-24445241 on Build GRCh38
907042 c.380C>T p.P127L Chr.1: 24445230-24445230 on Build GRCh38
6460315 c.371A>G p.K124R Chr.1: 24445221-24445221 on Build GRCh38
4833877 c.391C>T p.L131L Chr.1: 24445241-24445241 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs916530695 Chr.1: 24445311-24445311 on Build GRCh38
rs935423320 Chr.1: 24445315-24445315 on Build GRCh38
rs148899776 Chr.1: 24445302-24445302 on Build GRCh38
rs760613330 Chr.1: 24445252-24445252 on Build GRCh38
rs763970790 Chr.1: 24445253-24445253 on Build GRCh38
rs150134145 Chr.1: 24445116-24445116 on Build GRCh38
rs781550235 Chr.1: 24445292-24445292 on Build GRCh38
rs747589546 Chr.1: 24445208-24445208 on Build GRCh38
rs367712413 Chr.1: 24445267-24445267 on Build GRCh38
rs778713435 Chr.1: 24445156-24445156 on Build GRCh38
rs568912884 Chr.1: 24445146-24445146 on Build GRCh38
rs755470002 Chr.1: 24445289-24445289 on Build GRCh38
rs979927911 Chr.1: 24445132-24445132 on Build GRCh38
rs768594360 Chr.1: 24445209-24445209 on Build GRCh38
rs780702198 Chr.1: 24445197-24445197 on Build GRCh38
rs377434875 Chr.1: 24445288-24445288 on Build GRCh38
rs886553222 Chr.1: 24445176-24445176 on Build GRCh38
rs538772323 Chr.1: 24445114-24445114 on Build GRCh38
rs554482329 Chr.1: 24445163-24445163 on Build GRCh38
rs144375095 Chr.1: 24445231-24445231 on Build GRCh38
rs144926436 Chr.1: 24445186-24445186 on Build GRCh38
rs775387736 Chr.1: 24445247-24445247 on Build GRCh38
rs374362215 Chr.1: 24445127-24445127 on Build GRCh38
rs750759371 Chr.1: 24445158-24445158 on Build GRCh38
rs781282034 Chr.1: 24445218-24445218 on Build GRCh38
rs771896782 Chr.1: 24445240-24445240 on Build GRCh38
rs184039335 Chr.1: 24445147-24445147 on Build GRCh38
rs775690499 Chr.1: 24445230-24445230 on Build GRCh38
rs959794007 Chr.1: 24445159-24445159 on Build GRCh38
rs768864793 Chr.1: 24445270-24445270 on Build GRCh38
rs373918555 Chr.1: 24445221-24445221 on Build GRCh38
rs112029539 Chr.1: 24445285-24445285 on Build GRCh38
rs558350589 Chr.1: 24445255-24445255 on Build GRCh38
rs1041253455 Chr.1: 24445297-24445297 on Build GRCh38
rs761243406 Chr.1: 24445257-24445257 on Build GRCh38
rs199684969 Chr.1: 24445287-24445287 on Build GRCh38
rs534022144 Chr.1: 24445258-24445258 on Build GRCh38
rs758802459 Chr.1: 24445282-24445282 on Build GRCh38
rs760074920 Chr.1: 24445139-24445139 on Build GRCh38

View Full Product Details