Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 41810847-41811092 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAPKBP1
Entrez Gene ID
23005
Gene Name
mitogen-activated protein kinase binding protein 1
Gene Aliases
JNKBP-1, JNKBP1
UniGene
Hs.513661
Gene Chromosome Location
Chr.15: 41774434-41827855 on Build GRCh38
Transcripts
NM_001128608.1 NM_001265611.1 NM_014994.2 NR_049761.1 NR_049762.1 XM_006720438.1 XM_011521383.1 XM_011521384.2 XM_017022017.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4435404 c.207-3C>T p.? Chr.15: 41810880-41810880 on Build GRCh38
7150523 c.266C>T p.S89F Chr.15: 41810942-41810942 on Build GRCh38
4435403 c.207-3C>T p.? Chr.15: 41810880-41810880 on Build GRCh38
2270945 c.207-1G>A p.? Chr.15: 41810882-41810882 on Build GRCh38
7150524 c.266C>T p.S89F Chr.15: 41810942-41810942 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs540096426 Chr.15: 41810966-41810966 on Build GRCh38
rs762281420 Chr.15: 41810917-41810917 on Build GRCh38
rs757137844 Chr.15: 41810959-41810959 on Build GRCh38
rs749269704 Chr.15: 41810872-41810872 on Build GRCh38
rs746629282 Chr.15: 41810990-41810990 on Build GRCh38
rs980033996 Chr.15: 41811016-41811016 on Build GRCh38
rs201805153 Chr.15: 41810909-41810909 on Build GRCh38
rs199917154 Chr.15: 41810903-41810903 on Build GRCh38
rs757300762 Chr.15: 41810873-41810873 on Build GRCh38
rs200855888 Chr.15: 41810972-41810972 on Build GRCh38
rs773165079 Chr.15: 41810932-41810932 on Build GRCh38
rs370375890 Chr.15: 41810976-41810976 on Build GRCh38
rs376170248 Chr.15: 41810874-41810874 on Build GRCh38
rs764180765 Chr.15: 41810908-41810908 on Build GRCh38
rs373813643 Chr.15: 41810978-41810978 on Build GRCh38
rs547414773 Chr.15: 41810907-41810907 on Build GRCh38
rs778913039 Chr.15: 41810962-41810962 on Build GRCh38
rs765834202 Chr.15: 41810918-41810918 on Build GRCh38
rs763100511 Chr.15: 41810934-41810934 on Build GRCh38
rs763951461 Chr.15: 41810943-41810943 on Build GRCh38
rs778546981 Chr.15: 41810991-41810991 on Build GRCh38
rs775181068 Chr.15: 41810989-41810990 on Build GRCh38
rs375878673 Chr.15: 41810912-41810912 on Build GRCh38
rs956244504 Chr.15: 41811064-41811064 on Build GRCh38
rs969683992 Chr.15: 41811006-41811006 on Build GRCh38
rs200453027 Chr.15: 41810939-41810939 on Build GRCh38
rs753736254 Chr.15: 41810949-41810949 on Build GRCh38
rs928527620 Chr.15: 41811024-41811024 on Build GRCh38
rs745450007 Chr.15: 41810878-41810878 on Build GRCh38
rs766457904 Chr.15: 41810936-41810936 on Build GRCh38
rs751855057 Chr.15: 41810938-41810938 on Build GRCh38
rs1000088224 Chr.15: 41810961-41810961 on Build GRCh38

View Full Product Details