Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 22692594-22692831 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EGR3
Entrez Gene ID
1960
Gene Name
early growth response 3
Gene Aliases
EGR-3, PILOT
UniGene
Hs.534313
Gene Chromosome Location
Chr.8: 22687661-22693302 on Build GRCh38
Transcripts
NM_004430.2 XM_005273426.3 XM_011544429.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3412920 c.147G>A p.M49I Chr.8: 22692798-22692798 on Build GRCh38
3765398 c.144G>T p.Q48H Chr.8: 22692801-22692801 on Build GRCh38
3898984 c.137A>G p.Y46C Chr.8: 22692808-22692808 on Build GRCh38
5874388 c.133C>T p.H45Y Chr.8: 22692812-22692812 on Build GRCh38
2786426 c.131T>C p.V44A Chr.8: 22692814-22692814 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758184584 Chr.8: 22692730-22692732 on Build GRCh38
rs563377390 Chr.8: 22692620-22692620 on Build GRCh38
rs760221687 Chr.8: 22692812-22692812 on Build GRCh38
rs750229993 Chr.8: 22692743-22692743 on Build GRCh38
rs924863584 Chr.8: 22692720-22692720 on Build GRCh38
rs549089212 Chr.8: 22692676-22692676 on Build GRCh38
rs993038273 Chr.8: 22692768-22692768 on Build GRCh38
rs373059385 Chr.8: 22692771-22692771 on Build GRCh38
rs774799132 Chr.8: 22692801-22692801 on Build GRCh38
rs772426856 Chr.8: 22692767-22692768 on Build GRCh38
rs770267755 Chr.8: 22692770-22692770 on Build GRCh38
rs773470563 Chr.8: 22692775-22692776 on Build GRCh38
rs749799880 Chr.8: 22692783-22692783 on Build GRCh38
rs779907254 Chr.8: 22692753-22692753 on Build GRCh38
rs369825179 Chr.8: 22692761-22692761 on Build GRCh38
rs770711526 Chr.8: 22692676-22692677 on Build GRCh38
rs530892952 Chr.8: 22692668-22692668 on Build GRCh38
rs771244734 Chr.8: 22692794-22692794 on Build GRCh38
rs979436692 Chr.8: 22692707-22692707 on Build GRCh38
rs747163450 Chr.8: 22692760-22692760 on Build GRCh38
rs985835982 Chr.8: 22692644-22692644 on Build GRCh38
rs962197559 Chr.8: 22692673-22692673 on Build GRCh38
rs758210330 Chr.8: 22692747-22692747 on Build GRCh38
rs1016394270 Chr.8: 22692649-22692649 on Build GRCh38
rs968320739 Chr.8: 22692680-22692680 on Build GRCh38
rs748291085 Chr.8: 22692769-22692769 on Build GRCh38
rs567487553 Chr.8: 22692763-22692763 on Build GRCh38
rs972744719 Chr.8: 22692702-22692703 on Build GRCh38

View Full Product Details