Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41802844-41803095 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
P3H4
Entrez Gene ID
10609
Gene Name
prolyl 3-hydroxylase family member 4 (non-enzymatic)
Gene Aliases
LEPREL4, NO55, NOL55, SC65
UniGene
Hs.446459
Gene Chromosome Location
Chr.17: 41801952-41812617 on Build GRCh38
Transcripts
NM_006455.2 XM_006721640.3

SNP Detail

SNP ID Chromosome Location
rs782229779 Chr.17: 41802911-41802911 on Build GRCh38
rs940573400 Chr.17: 41802947-41802947 on Build GRCh38
rs782366640 Chr.17: 41802960-41802960 on Build GRCh38
rs368602485 Chr.17: 41803000-41803000 on Build GRCh38
rs372056957 Chr.17: 41803006-41803006 on Build GRCh38
rs1065029 Chr.17: 41802937-41802937 on Build GRCh38
rs782341621 Chr.17: 41802912-41802912 on Build GRCh38
rs921674268 Chr.17: 41802910-41802910 on Build GRCh38
rs984582026 Chr.17: 41802918-41802918 on Build GRCh38
rs73305750 Chr.17: 41803063-41803063 on Build GRCh38
rs201180524 Chr.17: 41802922-41802922 on Build GRCh38
rs782137328 Chr.17: 41802977-41802977 on Build GRCh38
rs373961470 Chr.17: 41802983-41802983 on Build GRCh38
rs781896114 Chr.17: 41802970-41802970 on Build GRCh38
rs1055814680 Chr.17: 41803047-41803047 on Build GRCh38
rs782598512 Chr.17: 41803020-41803020 on Build GRCh38
rs781789148 Chr.17: 41803017-41803017 on Build GRCh38
rs782054458 Chr.17: 41802945-41802945 on Build GRCh38
rs782404773 Chr.17: 41802943-41802943 on Build GRCh38
rs549251904 Chr.17: 41802887-41802887 on Build GRCh38
rs115439797 Chr.17: 41802913-41802913 on Build GRCh38
rs947576248 Chr.17: 41803077-41803077 on Build GRCh38
rs12451233 Chr.17: 41802875-41802875 on Build GRCh38
rs782759557 Chr.17: 41802964-41802964 on Build GRCh38
rs781941689 Chr.17: 41802944-41802944 on Build GRCh38
rs782225777 Chr.17: 41803064-41803064 on Build GRCh38
rs781867806 Chr.17: 41803027-41803027 on Build GRCh38
rs372964827 Chr.17: 41803002-41803002 on Build GRCh38

View Full Product Details