Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 55789178-55789449 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NLRP11
Entrez Gene ID
204801
Gene Name
NLR family pyrin domain containing 11
Gene Aliases
CLR19.6, NALP11, NOD17, PAN10, PYPAF6, PYPAF7
UniGene
Hs.375039
Gene Chromosome Location
Chr.19: 55785397-55836762 on Build GRCh38
Transcripts
NM_001297743.1 NM_145007.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2969498 c.2539G>A p.D847N Chr.19: 55789374-55789374 on Build GRCh38
5707217 c.2630T>C p.M877T Chr.19: 55789283-55789283 on Build GRCh38
5414553 c.2515C>T p.L839L Chr.19: 55789398-55789398 on Build GRCh38
317694 c.2683G>C p.G895R Chr.19: 55789230-55789230 on Build GRCh38
6152814 c.2589G>T p.R863S Chr.19: 55789324-55789324 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779653912 Chr.19: 55789373-55789373 on Build GRCh38
rs1000703185 Chr.19: 55789212-55789212 on Build GRCh38
rs1024246254 Chr.19: 55789341-55789352 on Build GRCh38
rs199960046 Chr.19: 55789423-55789423 on Build GRCh38
rs767152510 Chr.19: 55789280-55789280 on Build GRCh38
rs761682058 Chr.19: 55789384-55789384 on Build GRCh38
rs780891099 Chr.19: 55789239-55789239 on Build GRCh38
rs555849258 Chr.19: 55789203-55789203 on Build GRCh38
rs114044008 Chr.19: 55789375-55789375 on Build GRCh38
rs779546188 Chr.19: 55789315-55789315 on Build GRCh38
rs144599883 Chr.19: 55789390-55789390 on Build GRCh38
rs988689537 Chr.19: 55789330-55789338 on Build GRCh38
rs768550070 Chr.19: 55789267-55789267 on Build GRCh38
rs142033132 Chr.19: 55789374-55789374 on Build GRCh38
rs753175866 Chr.19: 55789341-55789341 on Build GRCh38
rs766514425 Chr.19: 55789304-55789304 on Build GRCh38
rs376135507 Chr.19: 55789232-55789232 on Build GRCh38
rs777939498 Chr.19: 55789353-55789353 on Build GRCh38
rs545829863 Chr.19: 55789202-55789202 on Build GRCh38
rs772846472 Chr.19: 55789422-55789422 on Build GRCh38
rs146254026 Chr.19: 55789378-55789378 on Build GRCh38
rs750672577 Chr.19: 55789286-55789286 on Build GRCh38
rs760762458 Chr.19: 55789287-55789287 on Build GRCh38
rs774812188 Chr.19: 55789248-55789248 on Build GRCh38
rs770595894 Chr.19: 55789392-55789395 on Build GRCh38
rs769358144 Chr.19: 55789245-55789245 on Build GRCh38
rs762908354 Chr.19: 55789259-55789259 on Build GRCh38
rs576092852 Chr.19: 55789222-55789222 on Build GRCh38
rs143293675 Chr.19: 55789253-55789253 on Build GRCh38
rs148312368 Chr.19: 55789307-55789307 on Build GRCh38
rs370176311 Chr.19: 55789264-55789264 on Build GRCh38
rs745443313 Chr.19: 55789242-55789242 on Build GRCh38
rs769289325 Chr.19: 55789364-55789364 on Build GRCh38
rs758835223 Chr.19: 55789342-55789342 on Build GRCh38
rs940607297 Chr.19: 55789405-55789405 on Build GRCh38
rs745555621 Chr.19: 55789363-55789363 on Build GRCh38
rs761593692 Chr.19: 55789273-55789273 on Build GRCh38
rs753941347 Chr.19: 55789305-55789305 on Build GRCh38

View Full Product Details