Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 152638296-152638531 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRQ
Entrez Gene ID
55879
Gene Name
gamma-aminobutyric acid type A receptor theta subunit
Gene Aliases
THETA
UniGene
Hs.283081
Gene Chromosome Location
Chr.X: 152637895-152659827 on Build GRCh38
Transcripts
NM_018558.3 XM_011531184.2

SNP Detail

SNP ID Chromosome Location
rs782011351 Chr.X: 152638374-152638374 on Build GRCh38
rs782354506 Chr.X: 152638387-152638387 on Build GRCh38
rs369828344 Chr.X: 152638458-152638458 on Build GRCh38
rs782576569 Chr.X: 152638447-152638447 on Build GRCh38
rs183563437 Chr.X: 152638348-152638348 on Build GRCh38
rs372073631 Chr.X: 152638364-152638364 on Build GRCh38
rs912695769 Chr.X: 152638457-152638457 on Build GRCh38
rs782412200 Chr.X: 152638368-152638368 on Build GRCh38
rs951657788 Chr.X: 152638399-152638399 on Build GRCh38
rs782619342 Chr.X: 152638324-152638324 on Build GRCh38
rs781956726 Chr.X: 152638390-152638390 on Build GRCh38
rs782201190 Chr.X: 152638325-152638325 on Build GRCh38
rs782256219 Chr.X: 152638379-152638379 on Build GRCh38
rs986975977 Chr.X: 152638433-152638433 on Build GRCh38

View Full Product Details