Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 109441410-109441667 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMPD2
Entrez Gene ID
6610
Gene Name
sphingomyelin phosphodiesterase 2
Gene Aliases
ISC1, NSMASE, NSMASE1
UniGene
Hs.55235
Gene Chromosome Location
Chr.6: 109440646-109443922 on Build GRCh38
Transcripts
NM_003080.2 XM_005267109.1 XM_011536079.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6423321 c.133G>A p.A45T Chr.6: 109441439-109441439 on Build GRCh38
4946526 c.116A>G p.Q39R Chr.6: 109441422-109441422 on Build GRCh38
3366250 c.177G>A p.L59L Chr.6: 109441581-109441581 on Build GRCh38
5807668 c.179G>T p.R60I Chr.6: 109441583-109441583 on Build GRCh38
3155238 c.159G>A p.E53E Chr.6: 109441563-109441563 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757169176 Chr.6: 109441436-109441436 on Build GRCh38
rs561839909 Chr.6: 109441498-109441498 on Build GRCh38
rs746449537 Chr.6: 109441617-109441617 on Build GRCh38
rs768173839 Chr.6: 109441584-109441585 on Build GRCh38
rs533969815 Chr.6: 109441590-109441590 on Build GRCh38
rs1048629622 Chr.6: 109441447-109441449 on Build GRCh38
rs758894027 Chr.6: 109441607-109441607 on Build GRCh38
rs937600664 Chr.6: 109441493-109441493 on Build GRCh38
rs746424969 Chr.6: 109441606-109441606 on Build GRCh38
rs556201093 Chr.6: 109441530-109441530 on Build GRCh38
rs895443299 Chr.6: 109441616-109441616 on Build GRCh38
rs776098031 Chr.6: 109441643-109441643 on Build GRCh38
rs765241160 Chr.6: 109441526-109441526 on Build GRCh38
rs779607068 Chr.6: 109441614-109441614 on Build GRCh38
rs186439077 Chr.6: 109441494-109441494 on Build GRCh38
rs749607058 Chr.6: 109441454-109441454 on Build GRCh38
rs778654994 Chr.6: 109441487-109441487 on Build GRCh38
rs745416319 Chr.6: 109441472-109441472 on Build GRCh38
rs569904970 Chr.6: 109441564-109441564 on Build GRCh38
rs779308551 Chr.6: 109441603-109441603 on Build GRCh38
rs146435492 Chr.6: 109441585-109441585 on Build GRCh38
rs189970558 Chr.6: 109441553-109441553 on Build GRCh38
rs756467144 Chr.6: 109441575-109441575 on Build GRCh38
rs761457883 Chr.6: 109441522-109441522 on Build GRCh38
rs772981231 Chr.6: 109441527-109441527 on Build GRCh38
rs768577877 Chr.6: 109441577-109441577 on Build GRCh38
rs779242483 Chr.6: 109441471-109441471 on Build GRCh38
rs762950702 Chr.6: 109441531-109441531 on Build GRCh38
rs373187364 Chr.6: 109441591-109441591 on Build GRCh38
rs767752447 Chr.6: 109441545-109441545 on Build GRCh38
rs768368587 Chr.6: 109441618-109441618 on Build GRCh38
rs375281708 Chr.6: 109441504-109441504 on Build GRCh38
rs906224392 Chr.6: 109441571-109441571 on Build GRCh38
rs771486808 Chr.6: 109441467-109441467 on Build GRCh38
rs752807798 Chr.6: 109441547-109441547 on Build GRCh38
rs776489782 Chr.6: 109441520-109441520 on Build GRCh38

View Full Product Details