Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44322619-44322888 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC25A39
Entrez Gene ID
51629
Gene Name
solute carrier family 25 member 39
Gene Aliases
CGI-69, CGI69
UniGene
Hs.744880
Gene Chromosome Location
Chr.17: 44319625-44324870 on Build GRCh38
Transcripts
NM_001143780.2 NM_001321240.1 NM_001321241.1 NM_016016.3 XM_011524880.2 XM_011524882.2 XM_011524883.2 XM_011524884.1 XM_017024737.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5662185 c.168G>A p.L56L Chr.17: 44322830-44322830 on Build GRCh38
3190979 c.170G>A p.W57* Chr.17: 44322828-44322828 on Build GRCh38
6037960 c.175C>A p.L59I Chr.17: 44322823-44322823 on Build GRCh38
5662186 c.168G>A p.L56L Chr.17: 44322830-44322830 on Build GRCh38
4267614 c.170G>A p.W57* Chr.17: 44322828-44322828 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs980916151 Chr.17: 44322690-44322690 on Build GRCh38
rs570886102 Chr.17: 44322661-44322661 on Build GRCh38
rs776242673 Chr.17: 44322786-44322786 on Build GRCh38
rs912804184 Chr.17: 44322707-44322707 on Build GRCh38
rs758289711 Chr.17: 44322832-44322832 on Build GRCh38
rs76073410 Chr.17: 44322789-44322789 on Build GRCh38
rs892365814 Chr.17: 44322787-44322787 on Build GRCh38
rs917931223 Chr.17: 44322682-44322682 on Build GRCh38
rs746892088 Chr.17: 44322784-44322784 on Build GRCh38
rs747989857 Chr.17: 44322765-44322765 on Build GRCh38
rs138244664 Chr.17: 44322812-44322812 on Build GRCh38
rs936520087 Chr.17: 44322772-44322772 on Build GRCh38
rs371734146 Chr.17: 44322691-44322691 on Build GRCh38
rs762199715 Chr.17: 44322791-44322791 on Build GRCh38
rs1032184559 Chr.17: 44322866-44322866 on Build GRCh38
rs1039870124 Chr.17: 44322715-44322715 on Build GRCh38
rs776416381 Chr.17: 44322859-44322859 on Build GRCh38
rs932976381 Chr.17: 44322698-44322698 on Build GRCh38
rs376488686 Chr.17: 44322818-44322818 on Build GRCh38
rs367929986 Chr.17: 44322836-44322836 on Build GRCh38
rs995613627 Chr.17: 44322749-44322749 on Build GRCh38
rs913599753 Chr.17: 44322673-44322673 on Build GRCh38
rs771926661 Chr.17: 44322775-44322775 on Build GRCh38
rs942928351 Chr.17: 44322709-44322709 on Build GRCh38
rs773312234 Chr.17: 44322776-44322776 on Build GRCh38
rs374272657 Chr.17: 44322853-44322853 on Build GRCh38
rs1051372108 Chr.17: 44322705-44322705 on Build GRCh38
rs758962039 Chr.17: 44322788-44322788 on Build GRCh38
rs766793744 Chr.17: 44322816-44322816 on Build GRCh38
rs767845153 Chr.17: 44322763-44322763 on Build GRCh38
rs948094588 Chr.17: 44322687-44322687 on Build GRCh38
rs898631347 Chr.17: 44322747-44322747 on Build GRCh38
rs371908215 Chr.17: 44322796-44322796 on Build GRCh38
rs988260837 Chr.17: 44322645-44322645 on Build GRCh38
rs779200449 Chr.17: 44322821-44322821 on Build GRCh38
rs532010769 Chr.17: 44322689-44322689 on Build GRCh38
rs372058439 Chr.17: 44322840-44322840 on Build GRCh38
rs750767670 Chr.17: 44322797-44322797 on Build GRCh38
rs992538530 Chr.17: 44322671-44322671 on Build GRCh38
rs746767242 Chr.17: 44322839-44322839 on Build GRCh38
rs925098154 Chr.17: 44322740-44322740 on Build GRCh38

View Full Product Details