Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 48706467-48706708 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SUV39H1
Entrez Gene ID
6839
Gene Name
suppressor of variegation 3-9 homolog 1
Gene Aliases
H3-K9-HMTase 1, KMT1A, MG44, SUV39H
UniGene
Hs.522639
Gene Chromosome Location
Chr.X: 48695554-48709016 on Build GRCh38
Transcripts
NM_001282166.1 NM_003173.3

SNP Detail

SNP ID Chromosome Location
rs782792759 Chr.X: 48706658-48706658 on Build GRCh38
rs782767775 Chr.X: 48706659-48706659 on Build GRCh38
rs782037864 Chr.X: 48706608-48706608 on Build GRCh38
rs782423798 Chr.X: 48706555-48706555 on Build GRCh38
rs781885078 Chr.X: 48706677-48706677 on Build GRCh38
rs781900909 Chr.X: 48706539-48706539 on Build GRCh38
rs782043171 Chr.X: 48706605-48706605 on Build GRCh38
rs79235204 Chr.X: 48706499-48706499 on Build GRCh38
rs973790599 Chr.X: 48706493-48706493 on Build GRCh38
rs781827609 Chr.X: 48706649-48706649 on Build GRCh38
rs782209222 Chr.X: 48706491-48706491 on Build GRCh38
rs372441780 Chr.X: 48706634-48706634 on Build GRCh38
rs782355758 Chr.X: 48706524-48706524 on Build GRCh38
rs1026744699 Chr.X: 48706575-48706575 on Build GRCh38
rs782132107 Chr.X: 48706545-48706545 on Build GRCh38
rs782715815 Chr.X: 48706631-48706631 on Build GRCh38
rs782057614 Chr.X: 48706617-48706617 on Build GRCh38

View Full Product Details